The reference solutions are contained in colorless neutral glass grade tubes. These are manufactured to meet the specifications for Type I borosilicate glass, which is the relevant Ph. Eur. standard for containers of this nature. The set is specifically developed for Method 1 as described in Ph. Eur. chapter 2.2.2 "Degree of coloration of liquids." Unfortunately, the dimensions of the ampoules are not specified, as Ph. Eur. chapter 2.2.2 does not require them to be specified.
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
analytical standard
Quality Segment
agency
according to Ph. Eur. color intensity testing B1-B9
product line
Certipur®
application(s)
coatings
environmental
food and beverages
pharmaceutical
format
multi-component solution
storage temp.
15-25°C
General description
In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website. Download your certificate at https://www.sigmaaldrich.com/US/en/documents-search to view certified values, including uncertainty and expiry date.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| format multi-component solution | format multi-component solution | format multi-component solution | format multi-component solution |
| product line Certipur® | product line Certipur® | product line - | product line - |
| grade analytical standard | grade analytical standard | grade analytical standard | grade analytical standard |
| application(s) coatings | application(s) coatings | application(s) cleaning products | application(s) cleaning products |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. - | storage temp. - |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
signalword
Danger
Hazard Classifications
Aquatic Chronic 2 - Carc. 1B - Met. Corr. 1 - Repr. 1B
Storage Class
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Could you please confirm whether the 10 mL glass ampoules used for your Colour Reference Solutions B (Ph. Eur., B1–B9) comply with the relevant Ph. Eur. requirements for glass containers?Additionally, could you provide the dimensions of the ampoules.
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdfHelpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-






