Skip to Content
Merck

Skip To

M-147

Methedrone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

4-Methoxymethcathinone hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C11H16ClNO2
CAS Number:
Molecular Weight:
229.70
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CNC(C)C(=O)c1ccc(OC)cc1

InChI

1S/C11H15NO2.ClH/c1-8(12-2)11(13)9-4-6-10(14-3)7-5-9;/h4-8,12H,1-3H3;1H

InChI key

QIJFAKGYWIXUDG-UHFFFAOYSA-N

General description

Methedrone is a synthetic stimulant of the phenethylamine, amphetamine, and cathinone classes. Significant abuse of this new designer drug, also known as 4-methoxymeth-cathinone, methoxyphedrine, or bk-PMMA, has been reported within the last year in Sweden, the UK, Poland and states including Louisiana and Kentucky.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M-138M-140M-141
application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

format

single component solution

format

single component solution

format

single component solution

format

single component solution

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters