Skip to Content
Merck

Skip To

M-136

Methotrexate solution

1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H22N8O5
CAS Number:
Molecular Weight:
454.44
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol with 0.1N NaOH

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=CC=C(N(C)CC2=NC3=C(N=C2)N=C(N)N=C3N)C=C1)NC(C(O)=O)CCC(O)=O

InChI

1S/C20H22N8O5/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27)

InChI key

FBOZXECLQNJBKD-UHFFFAOYSA-N

General description

A Certified Spiking Solution® for use in therapeutic drug monitoring (TDM) analyses by HPLC or LC/MS. Used in the treatment of cancer and autoimmune diseases, methotrexate is an antimetabolite and antifolate drug monitored by clinical labs to ensure patients remain within the drug′s therapeutic range. The Snap-N-Spike® format allows laboratories to eliminate weighing operations of hazardous substances such as methotrexate, significantly reducing occupational exposure hazards related to handling of acutely toxic powders, and provides the ability to spike into their matrix of choice just before use.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR13961414003M1000000
application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

form

liquid

form

-

form

-

form

-

Quality Level

300

Quality Level

300

Quality Level

-

Quality Level

-

grade

certified reference material

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Joshua F Baker et al.
Annals of the rheumatic diseases, 73(11), 1968-1974 (2013-08-02)
To determine if early MRI measures predict X-ray progression at 1 and 2 years in a large RA trial cohort. This study included 256 methotrexate (MTX)-naïve RA patients from a randomised placebo-controlled trial of golimumab (GO-BEFORE). MRIs of wrist and 2nd-5th
J Westra et al.
Clinical and experimental immunology, 178(1), 40-47 (2014-06-04)
Rituximab (RTX) treatment in rheumatoid arthritis (RA) patients severely hampers humoral response after influenza vaccination as determined by haemagglutination inhibition assay (HI). It is not known whether HI reflects both immunoglobulin (Ig)M and IgG (subclass) influenza response, and whether IgM
Josef S Smolen et al.
Lancet (London, England), 383(9914), 321-332 (2013-10-31)
Biological agents offer good control of rheumatoid arthritis, but the long-term benefits of achieving low disease activity with a biological agent plus methotrexate or methotrexate alone are unclear. The OPTIMA trial assessed different treatment adjustment strategies in patients with early



Global Trade Item Number

SKUGTIN
M-136-1ML04061833977071

Questions

Reviews

No rating value

Active Filters