Skip to Content
Merck

Skip To

E-015

Ethyl-β-D-glucuronide

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H14O7
CAS Number:
Molecular Weight:
222.19
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O)[C@@H](C(O)=O)O[C@@H](OCC)[C@@H]1O

InChI

1S/C8H14O7/c1-2-14-8-5(11)3(9)4(10)6(15-8)7(12)13/h3-6,8-11H,2H2,1H3,(H,12,13)/t3-,4-,5+,6-,8+/m0/s1

InChI key

IWJBVMJWSPZNJH-UQGZVRACSA-N

General description

This Certified Reference Material is widely used in forensic analysis and clinical toxicology applications to determine blood alcohol content.

Ethyl-β-D-glucuronide (EtG) is a water-soluble and stable metabolite of ethanol, used as a routine biomarker in alcohol abuse monitoring. A very small fraction of ethanol undergoes degradation to EtG, and it can be detected in body fluids as well as hair.

Application

The certified reference solution (CRM) in solution can also be used as follows:
  • Multi-residue analysis of different alcohol metabolites in human blood plasma and urine samples by ultra-high performance liquid chromatography (UHPLC) combined with tandem mass spectrometry (MS/MS)
  • Optimization of a supercritical fluid chromatography-tandem mass spectrometric (SFC-MS/MS) method for the separation and quantification of highly polar drugs and their metabolites using an ion-exchange stationary phase from human urine samples
  • Evaluation of a paired ion electrospray ionization mass spectrometric (PIESI-MS) approach coupled with high performance liquid chromatographic (HPLC) procedure for the detection and measurement of drug metabolites in urine samples
  • Assess the impact of sample preparation of hair samples on the quantitative analysis of ethyl-β-D-glucuronide by ultra-high performance liquid chromatography-electrospray ionization-tandem mass spectrometry[1]
  • Analysis of ethyl-β-D-glucuronide as a urinary biomarker in human urine samples by a benchtop gas chromatograph coupled with a mass spectrometer (GC-MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E-048E-016E-063
application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

format

single component solution

format

single component solution

format

single component solution

format

single component solution

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material


Still not finding the right product?

Explore all of our products under Ethyl-β-D-glucuronide


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sensitive detection of anionic metabolites of drugs by positive ion mode HPLC-PIESI-MS
Guo H, et al.
International Journal of Mass Spectrometry, 389, 14-25 (2015)
A simplified approach for determination of urinary ethyl glucuronide by gas chromatography-mass spectrometry
Ghosh S, et al.
Journal of analytical science and technology, 12, 1-9 (2021)
A versatile UHPLC-MSMS method for simultaneous quantification of various alcohol intake related compounds in human urine and blood
Monosik R and Dragsted LO
Analytical Methods : Advancing Methods and Applications, 8, 6865-6871 (2016)



Global Trade Item Number

SKUGTIN
E-015-1ML04061838055040

Questions

Reviews

No rating value

Active Filters