Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Level
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
clinical testing
format
single component solution
storage temp.
−20°C
SMILES string
ClC1=CC2=C(C=C1)CCC3=C(C=CC=C3)N2CCCNC([2H])([2H])[2H].[H]Cl
InChI
1S/C18H21ClN2.ClH/c1-20-11-4-12-21-17-6-3-2-5-14(17)7-8-15-9-10-16(19)13-18(15)21;/h2-3,5-6,9-10,13,20H,4,7-8,11-12H2,1H3;1H/i1D3;
InChI key
KMDDAZOLOSKTKZ-NIIDSAIPSA-N
General description
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| format single component solution | format single component solution | format single component solution | format single component solution |
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| concentration 100 μg/mL in methanol (as free base) | concentration 100 μg/mL in methanol (as free base) | concentration 1.0 mg/mL in methanol (as free base) | concentration 100 μg/mL in acetonitrile |
| manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® |
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| D-113-1ML | 04061833557174 |





