Skip to Content
Merck

Skip To

D-102

N,N-Dimethyltryptamine (DMT) solution

1.0 mg/mL in methanol, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H16N2
CAS Number:
Molecular Weight:
188.27
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN(C)CCC1=CNC2=CC=CC=C21

InChI

1S/C12H16N2/c1-14(2)8-7-10-9-13-12-6-4-3-5-11(10)12/h3-6,9,13H,7-8H2,1-2H3

InChI key

DMULVCHRPCFFGV-UHFFFAOYSA-N

General description

This certified solution standard is suitable for N,N-Dimethyltryptamine (DMT) testing methods by GC/MS or LC/MS in clinical toxicology, urine drug testing, or forensic analysis applications. DMT is a naturally occurring hallucinogenic tryptamine derived from the amino acid tryptophan. Abuse of DMT reportedly can cause auditory and visual hallucinations, and mild stimulant effects such as increased heart rate and blood pressure.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M-168D-017P-098
grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

format

single component solution

format

single component solution

format

single component solution

format

single component solution

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−70°C

form

liquid

form

liquid

form

liquid

form

liquid

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

pharmaceutical (small molecule)

application(s)

forensics and toxicology

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

(Snap-N-Spike®)


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Is it possible to request this substance in an ethanol solution? We need it for IV administration.

    1 answer
    1. This product is intended for use as an analytical standard.

      Preparing this compound in ethanol will require a custom order. Please navigate to the link below. Scroll down to fill out and submit the request form. A member of the custom sales team will respond shortly.
      https://www.sigmaaldrich.com/services/custom-products/reference-materials#form

      Helpful?

Reviews

No rating value

Active Filters