Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Level
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
20 μg/mL in methanol
format
single component solution
storage temp.
−20°C
SMILES string
O1[C@H](CC(=C(C1=O)CO)C)[C@H]([C@@H]2[C@@]3([C@H]([C@@H]4[C@@H]5O[C@@H]5[C@@]6([C@@]([C@H]4CC3)(C(=O)C=CC6)C)O)CC2)C)C
InChI
1S/C28H38O6/c1-14-12-20(33-25(31)16(14)13-29)15(2)17-7-8-18-22-19(9-11-26(17,18)3)27(4)21(30)6-5-10-28(27,32)24-23(22)34-24/h5-6,15,17-20,22-24,29,32H,7-13H2,1-4H3/t15-,17+,18-,19-,20+,22-,23-,24-,26+,27-,28-/m0/s1
InChI key
AWVMHXZWAKRDGG-MEBIVHGNSA-N
General description
Discover the entire range of phytochemical certified reference material solutions from Cerilliant
Other Notes
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F
flash_point_c
9.7 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A6426-500G | 04061835542376 |
| A6426-100G | 04061835542369 |
| A6426-1KG | 04061833382240 |
| A8311-50ML | 04061837896033 |
| MAB748-MG-K | 04053252747991 |
| BLED-1 | 04065270781666 |
| 21792 | 04061833370469 |
| SRP2140-10UG | 04061837201547 |
| SAB4301416-100UL | 04061837438745 |
| SAB4300195-100UG | 04061831496277 |
| SAB4504210-100UG | 04061832524405 |
| MAB1160 | 04053252265297 |
| RAB0350-1KT | 04061834961857 |
| RAB0348-1KT | 04061834961833 |
| RAB0349-1KT | 04061834961840 |
| E7028-1VL | 04061826590843 |
| RAB0352-1KT | 04061834961864 |
| A-204-1ML | 04065267915098 |
| HLTUD0358 | 04061836098759 |
| 22039 | 04061833028308 |
| 22013 | 04061833028063 |


