Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 0.5 mL
manufacturer/tradename
Cerilliant®
drug control
estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal), kontrollierte Droge in Deutschland
concentration
100 μg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
O=C(N(C1CCN(CC1)CCC2=CC=CC=C2)C3=CC=CC=C3)C=C.Cl
InChI key
YUHNRLIKLNYPDD-UHFFFAOYSA-N
General description
Legal Information
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.
English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
Disclaimer
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


