Skip to Content
Merck

Skip To

A-096

Aldosterone solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Aldosterone, 11β,21-Dihydroxy-3,20-dioxo-4-pregnen-18-al, 11β,21-Dihydroxypregn-4-ene-3,18,20-trione, 18-Aldocorticosterone, Reichstein X

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
200-835-2
Beilstein/REAXYS Number:
3224996
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@]4(C[C@H](O)[C@H]23)C=O

InChI

1S/C21H28O5/c1-20-7-6-13(24)8-12(20)2-3-14-15-4-5-16(18(26)10-22)21(15,11-23)9-17(25)19(14)20/h8,11,14-17,19,22,25H,2-7,9-10H2,1H3/t14-,15-,16+,17-,19+,20-,21+/m0/s1

InChI key

PQSUYGKTWSAVDQ-ZVIOFETBSA-N

General description

Aldosterone is a hormone in the mineralocorticoid family which stimulates sodium transport across cell membranes and assists in the maintenance of blood pressure and blood volume. Aldosterone levels in serum/plasma and urine are measured by LC-MS/MS for investigation of primary or secondary aldosteronism ranging from adrenal cortical hyperplasia to renovascular disease and Bartter′s syndrome.

Application

The d-isomer of aldosterone is considered to be the biologically active isomer.

Other Notes

Exists as an equilibrium mixture of the aldehyde and the hemiacetal.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9477F612746148
technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

form

liquid

form

powder

form

powder

form

-

format

single component solution

format

-

format

-

format

neat

Quality Level

300

Quality Level

200

Quality Level

100

Quality Level

100

grade

certified reference material

grade

-

grade

-

grade

analytical standard

storage temp.

−20°C

storage temp.

room temp

storage temp.

room temp

storage temp.

-


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Athina Markou et al.
The Journal of clinical endocrinology and metabolism, 98(4), 1409-1416 (2013-03-09)
Primary aldosteronism (PA) is an established cause of hypertension, whereas high-normal serum aldosterone levels have been linked to an increased risk for hypertension. We aimed to define the post-fludrocortisone-dexamethasone suppression test (FDST) normal cutoff values of aldosterone and the aldosterone
Baojian Xue et al.
Hypertension (Dallas, Tex. : 1979), 61(6), 1255-1262 (2013-04-24)
The identification of the specific estrogen receptor (ER) subtypes that are involved in estrogen protection from hypertension and their specific locations in the central nervous system is critical to our understanding and design of effective estrogen replacement therapies in women.
Felix Beuschlein
European journal of endocrinology, 168(6), R85-R93 (2013-04-10)
Arterial hypertension is a major cardiovascular risk factor that affects between 10 and 40% of the population in industrialized countries. Primary aldosteronism (PA) is the most common form of secondary hypertension with an estimated prevalence of around 10% in referral



Global Trade Item Number

SKUGTIN
A-096-1ML04061833326336

Questions

Reviews

No rating value

Active Filters