Skip to Content
Merck

Skip To

A-003

6-Acetylmorphine, 100 μg/mL solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

6-Acetylmorphine solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-835-2
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4OC(C)=O)c35

InChI

1S/C19H21NO4/c1-10(21)23-15-6-4-12-13-9-11-3-5-14(22)17-16(11)19(12,18(15)24-17)7-8-20(13)2/h3-6,12-13,15,18,22H,7-9H2,1-2H3/t12-,13+,15-,18-,19-/m0/s1

InChI key

JJGYGPZNTOPXGV-SSTWWWIQSA-N

General description

6-Acetylmorphine, also known as 6-monoacetylmorphine or 6-MAM, is an active metabolite of heroin and biomarker for heroin usage. Certified Spiking Solutions® are suitable for use as a starting material in calibrators and controls for GC/MS or LC/MS methods in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTIONS is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A-009A-006A-010
application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



M Pravetoni et al.
Vaccine, 30(31), 4617-4624 (2012-05-16)
Opioid conjugate vaccines have shown promise in animal models as a potential treatment for opioid addiction. Individual vaccines are quite specific and each targets only a limited number of structurally similar opioids. Since opioid users can switch or transition between
Vesna Milovanović et al.
Vojnosanitetski pregled, 69(2), 141-146 (2012-04-17)
Saliva represents an alternative specimen for substances abuse determination in toxicology. Hence, the aim of this study was to optimize a method for saliva specimen preparation for heroin metabolites, morphine and 6-monoacetylmorphine (6-mam), and codeine determination by liquid chromatography-mass spectrometry
Cai Tie et al.
Journal of mass spectrometry : JMS, 47(11), 1429-1434 (2012-11-14)
With the combination of high separation ability of capillary electrophoresis (CE) and strong identification ability of mass spectrometry (MS), CE/MS is becoming a powerful tool for polar and ionic analytes analysis. Different interfaces have been developed to enhance the sensitivity



Global Trade Item Number

SKUGTIN
1445595-2MG04065267682679
5465804061833457405
NIST839204065265448444
A-003-1ML04061833325919

Questions

Reviews

No rating value

Active Filters