If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product Name
Molybdenum(IV) sulfide, powder
form
powder
composition
MoS₂
reaction suitability
core: molybdenum
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
particle size
~6 μm (max. 40 μm)
density
5.06 g/mL at 25 °C (lit.)
greener alternative category
SMILES string
S=[Mo]=S
InChI
1S/Mo.2S
InChI key
CWQXQMHSOZUFJS-UHFFFAOYSA-N
General description
Application
1 of 1
This Item | |||
|---|---|---|---|
| particle size ~6 μm (max. 40 μm) | particle size - | particle size ≤500 nm | particle size - |
| form powder | form nanopowder | form dispersion | form - |
| density 5.06 g/mL at 25 °C (lit.) | density 5.06 g/mL at 25 °C (lit.) | density 0.9-1.1 g/mL at 25 °C | density 5.06 g/mL at 25 °C (lit.) |
Still not finding the right product?
Explore all of our products under Molybdenum(IV) sulfide
Storage Class
11 - Combustible Solids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 69860-100G | 04061832794198 |
| 69860-500G | 04061832794204 |
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-

