Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
grade
pharmaceutical primary standard
API family
titanium dioxide
manufacturer/tradename
USP
mp
>350 °C (lit.)
density
4.26 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
O=[Ti]=O
InChI
1S/2O.Ti
InChI key
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Titanium dioxide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename - |
| grade pharmaceutical primary standard | grade - | grade - | grade - |
| format neat | format - | format - | format - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) battery manufacturing | application(s) - |
| density 4.26 g/mL at 25 °C (lit.) | density - | density - | density - |
| mp >350 °C (lit.) | mp 1855 °C | mp 1843 °C | mp 1855 °C |
Still not finding the right product?
Explore all of our products under Titanium dioxide
Storage Class
11 - Combustible Solids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1667585-2G | 04061833861523 |
