Skip to Content
Merck

Skip To

1613804

USP

Sodium chloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Halite, NaCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3534976
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

vapor pressure

1 mmHg ( 865 °C)

API family

sodium chloride

manufacturer/tradename

USP

availability

not available in (Sales restrictions may apply)

pH

6.7-7.3

mp

801 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium chloride USP reference standard for use in specified quality tests and assays.

Additionally used to make standard and system suitability solutions for the assay and impurity analysis using Liquid chromatography in conjunction with Conductivity with a suppression detector according to United States Pharmacopeia (USP) Monographs :
  • Sodium Chloride tablets
  • Sodium Chloride Inhalation Solution
  • Sodium Chloride Ophthalmic Ointment
  • Sodium Chloride Ophthalmic Solution
  • Sodium Chloride Tablets for Solution
  • SodiumFluoride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR13211.06400S1679
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

liquid formulation
ophthalmics
pharmaceutical

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

-

format

-

mp

801 °C (lit.)

mp

801 °C (lit.)

mp

801 °C (lit.)

mp

801 °C (lit.)

pH

6.7-7.3

pH

-

pH

4.5-7.0 (20 °C, 100 g/L in H2O)

pH

7


Still not finding the right product?

Explore all of our products under Sodium chloride


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sodium Chloride Tablets for Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(1) (2021)
Sodium Chloride Ophthalmic Ointment
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(5) (2022)
Sodium Chloride Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(1) (2021)



Global Trade Item Number

SKUGTIN
1613804-1G04061838746313

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the shelf life of an unopened container of USP grade sodium chloride? Thanks

    1 answer
    1. This is a product of the United States Pharmacopeia. An expiration date is not assigned to this material. Please see the link below to review the USP Certificate of Analysis for the current lot:
      https://static.usp.org/pdf/EN/referenceStandards/certificates/1613804-R161V0.pdf

      Please see the link below to review the USP product dating policy:
      https://www.usp.org/sites/default/files/usp/document/our-work/reference-standards/usp-policy-statement-expiration-dating.pdf

      Helpful?

Reviews

No rating value

Active Filters