Skip to Content
Merck

Skip To

1605500

USP

Ropivacaine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ropivacaine hydrochloride monohydrate, (2S)-N-(2,6-dimethylphenyl)-1-propyl-2-piperidinecarboxamide hydrochloride monohydrate, (S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride monohydrate, Naropin hydrochloride monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H26N2O · HCl · H2O
CAS Number:
Molecular Weight:
328.88
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

ropivacaine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.Cl.CCCN1CCCC[C@H]1C(=O)Nc2c(C)cccc2C

InChI

1S/C17H26N2O.ClH.H2O/c1-4-11-19-12-6-5-10-15(19)17(20)18-16-13(2)8-7-9-14(16)3;;/h7-9,15H,4-6,10-12H2,1-3H3,(H,18,20);1H;1H2/t15-;;/m0../s1

InChI key

VSHFRHVKMYGBJL-CKUXDGONSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ropivacaine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fentanyl Citrate and Ropivacaine Hydrochloride Compounded Injection
  • Ropivacaine Hydrochloride
  • Ropivacaine Hydrochloride Injection

Biochem/physiol Actions

Local anaesthetic with less cardiotoxicity than bupivacaine.
Local anaesthetic with less cardiotoxicity than bupivacaine; causes reversible blockade of impulse propagation along nerve fibres by preventing the inward movement of sodium ions through the cell membrane of the nerve fibers.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2303BP699M0370000
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

BP

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

neat

format

neat

API family

ropivacaine

API family

ropivacaine

API family

mepivacaine

API family

mepivacaine


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1605500-200MG04061838745668

Questions

Reviews

No rating value

Active Filters