Skip to Content
Merck

Skip To

1437202

USP

Methylpyrrolidone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Methyl-2-pyrrolidinone, 1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
106420
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C), 0.99 mmHg ( 40 °C)

form

liquid

autoignition temp.

518 °F

expl. lim.

9.5 %

manufacturer/tradename

USP

refractive index

n20/D 1.47 (lit.)

bp

202 °C (lit.), 81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

General description

Methylpyrrolidone is a highly polar solvent used in biologics formulations, particularly for drugs that require solubilization of hydrophobic compounds. It facilitates the delivery and stability of poorly soluble biologics by enhancing their dissolution properties. NMP is widely used in drug delivery systems, contributing to more efficient therapeutic delivery.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylpyrrolidone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pha

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1352787691.02497
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

analytical standard

grade

-

format

neat

format

neat

format

neat

format

-

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

-

form

liquid

form

-

form

-

form

liquid

density

1.028 g/mL at 25 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

density

1.03 g/cm3 at 20 °C


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

195.8 °F - Pensky-Martens closed cup

flash_point_c

91 °C - Pensky-Martens closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ryo Matsumoto et al.
Scientific reports, 5, 12884-12884 (2015-08-11)
Peptides, especially intracellular functional peptides that can play a particular role inside a cell, have attracted attention as promising materials to control cell fate. However, hydrophilic materials like peptides are difficult for cells to internalize. Therefore, the screening and design



Global Trade Item Number

SKUGTIN
1437202-2G04061838735058

Questions

Reviews

No rating value

Active Filters