Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
API family
losartan
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
Clc1nc([n](c1COC(=O)C)Cc2ccc(cc2)c3c(cccc3)c4n[nH]nn4)CCCC
InChI
1S/C24H25ClN6O2/c1-3-4-9-22-26-23(25)21(15-33-16(2)32)31(22)14-17-10-12-18(13-11-17)19-7-5-6-8-20(19)24-27-29-30-28-24/h5-8,10-13H,3-4,9,14-15H2,1-2H3,(H,27,28,29,30)
InChI key
HIUYRCVDCVFSBP-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| format neat | format - | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C | storage temp. 2-8°C |
| API family losartan | API family losartan | API family losartan | API family losartan |
Still not finding the right product?
Explore all of our products under Losartan Related Compound B
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1370484-20MG | 04061833845264 |

