Skip to Content
Merck

Skip To

1218016

USP

Diphenhydramine Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(Diphenylmethoxy)-N-methylethanamine hydrochloride, N-Desmethyldiphenhydramine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H19NO · HCl
CAS Number:
Molecular Weight:
277.79
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

diphenhydramine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].[N+H2](CCOC(c2ccccc2)c1ccccc1)C

InChI

1S/C16H19NO.ClH/c1-17-12-13-18-16(14-8-4-2-5-9-14)15-10-6-3-7-11-15;/h2-11,16-17H,12-13H2,1H3;1H

InChI key

OARUOQLHKTZEIM-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diphenhydramine Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Diphenhydramine Hydrochloride Oral Powder
  • Diphenhydramine Citrate and Ibuprofen Tablets
  • Diphenhydramine Hydrochloride
  • Diphenhydramine Citrate
  • Diphenhydramine Hydrochloride Injection
  • Diphenhydramine Hydrochloride Capsules
  • Diphenhydramine Hydrochloride Oral Solution

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1899PHR19931218027
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

format

neat

format

-

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

API family

diphenhydramine

API family

diphenhydramine

API family

diphenhydramine

API family

diphenhydramine


Still not finding the right product?

Explore all of our products under Diphenhydramine Related Compound A


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1218016-25MG04061838719706

Questions

Reviews

No rating value

Active Filters