Skip to Content
Merck

Skip To

1154558

USP

Alpha Cyclodextrin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

α-Cyclodextrin, α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
79627
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

description

API family: alfadex

API family

cyclodextrin, alfadex

manufacturer/tradename

USP

mp

>278 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C36H60O30/c37-1-7-25-13(43)19(49)31(55-7)62-26-8(2-38)57-33(21(51)15(26)45)64-28-10(4-40)59-35(23(53)17(28)47)66-30-12(6-42)60-36(24(54)18(30)48)65-29-11(5-41)58-34(22(52)16(29)46)63-27-9(3-39)56-32(61-25)20(50)14(27)44/h7-54H,1-6H2/t7-,8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-/m1/s1

InChI key

HFHDHCJBZVLPGP-RWMJIURBSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alpha Cyclodextrin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C468077878828705
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

Wacker Chemie AG

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

-

grade

-

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

-

format

neat

format

-

format

-

format

-

API family

cyclodextrin, alfadex

API family

-

API family

-

API family

-

description

API family: alfadex

description

-

description

-

description

-


Still not finding the right product?

Explore all of our products under Alpha Cyclodextrin


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hidetoshi Arima
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 124(7), 451-464 (2004-07-06)
Due to the growing concerns over the toxicity and immunogenicity of viral DNA delivery systems, DNA delivery via nonviral routes has become more desirable and advantageous. In particular, polycation complexes with DNA (polyplex) are attractive nonviral vectors. To design novel
Martha Mullally et al.
Canadian journal of physiology and pharmacology, 92(9), 758-764 (2014-08-21)
The mode of action of the anxiolytic medicinal plant Souroubea sympetala was investigated to test the hypothesis that extracts and the active principle act at the pharmacologically important GABAA-benzodiazepine (GABAA-BZD) receptor. Leaf extracts prepared by ethyl acetate extraction or supercritical



Global Trade Item Number

SKUGTIN
1154558-500MG04061833859711

Questions

Reviews

No rating value

Active Filters