Skip to Content
Merck

Skip To

1065480

USP

Beta Carotene

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

β-Carotene, β,β-Carotene, Provitamin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1917416
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

carotenes

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C=C/C2=C(C)CCCC2(C)C)C(C)(C)CCC1

InChI

1S/C40H56/c1-31(19-13-21-33(3)25-27-37-35(5)23-15-29-39(37,7)8)17-11-12-18-32(2)20-14-22-34(4)26-28-38-36(6)24-16-30-40(38,9)10/h11-14,17-22,25-28H,15-16,23-24,29-30H2,1-10H3/b12-11+,19-13+,20-14+,27-25+,28-26+,31-17+,32-18+,33-21+,34-22+

InChI key

OENHQHLEOONYIE-JLTXGRSLSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Beta Carotene USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Beta Carotene Capsules
  • Beta Carotene Preparation
  • Carotenes
  • Oil-Soluble Vitamins Oral Solution
  • Spirulina Tablets
  • Spirulina
  • Oil-Soluble Vitamins with Minerals Oral Solution

Biochem/physiol Actions

The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro. However, in vivo it appears to act either as an antioxidant or a prooxidant depending on cellular environment. It reduces the incidence of many cancers, but enhances lung cancer incidence in smokers.
The most important of the provitamins A.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0002073PHR1239V-058
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

Cerilliant®

grade

pharmaceutical primary standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

food and beverages

application(s)

clinical testing

format

neat

format

-

format

neat

format

single component solution

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

−70°C

API family

carotenes

API family

-

API family

carotenes

API family

-


Still not finding the right product?

Explore all of our products under Beta Carotene


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

217.9 °F - closed cup

flash_point_c

103.3 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Leonardo M de Souza Mesquita et al.
Molecules (Basel, Switzerland), 26(13) (2021-07-03)
Currently, on an industrial scale, synthetic colorants are used in many fields, as well as those extracted with conventional organic solvents (COSs), leading to several environmental issues. Therefore, we developed a sustainable extraction and purification method mediated by ionic liquids



Global Trade Item Number

SKUGTIN
1065480-7X100MG04061838674166

Questions

Reviews

No rating value

Active Filters