Skip to Content
Merck

Skip To

1045574

USP

Azacitidine Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Amidino-3-β-D-ribofuranosylurea monopicrate, 1-β-D-Ribofuranosyl-3-guanylurea picrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H14N4O5 · C6H3N3O7
CAS Number:
Molecular Weight:
463.31
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

azacitidine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[N+](=O)([O-])c2c(c(cc(c2)[N+](=O)[O-])[N+](=O)[O-])O.N(C1OC(C(C1O)O)CO)C(=O)N=C(N)N

InChI

1S/C7H14N4O5.C6H3N3O7/c8-6(9)11-7(15)10-5-4(14)3(13)2(1-12)16-5;10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h2-5,12-14H,1H2,(H5,8,9,10,11,15);1-2,10H

InChI key

RTONSOUBEUPEOS-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Azacitidine Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
104553910455411090717
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

API family

azacitidine

API family

azacitidine

API family

azacitidine

API family

capecitabine


pictograms

Exploding BombSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Expl. 1.1

Storage Class

1 - Explosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1045574-15MG04061833840627

Questions

Reviews

No rating value

Active Filters