Skip to Content
Merck

Skip To

V8376

Val-Tyr-Val

≥98% (TLC), suitable for HPLC

Synonym(s):

L-valyl-L-tyrosyl-L-valine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H29N3O5
CAS Number:
Molecular Weight:
379.45
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Val-Tyr-Val, crystalline

assay

≥98% (TLC)

Quality Segment

form

crystalline

technique(s)

HPLC: suitable

color

off-white to tan

storage temp.

−20°C

SMILES string

CC(C)C(N)C(=O)NC(Cc1ccc(O)cc1)C(=O)NC(C(C)C)C(O)=O

InChI

1S/C19H29N3O5/c1-10(2)15(20)18(25)21-14(9-12-5-7-13(23)8-6-12)17(24)22-16(11(3)4)19(26)27/h5-8,10-11,14-16,23H,9,20H2,1-4H3,(H,21,25)(H,22,24)(H,26,27)

InChI key

ZNGPROMGGGFOAA-UHFFFAOYSA-N

Application

Val-Tyr-Val has been used as a constituent of high performance liquid chromatography (HPLC) standard protein mixture.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
47638V0882SML1472
form

crystalline

form

solid

form

powder

form

film

technique(s)

HPLC: suitable

technique(s)

-

technique(s)

-

technique(s)

-

assay

≥98% (TLC)

assay

≥98.0% (HPLC)

assay

≥98% (TLC)

assay

≥95% (HPLC)

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

color

off-white to tan

color

-

color

-

color

colorless

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
V8376-100MG04061833276518

Questions

Reviews

No rating value

Active Filters