Skip to Content
Merck

Skip To

T5293

Monoclonal Anti-β-Tubulin antibody produced in mouse

clone 2-28-33, ascites fluid

Synonym(s):

Anti-β-Tubulin Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
2-28-33, monoclonal
Application:
IF, WB
Citations:
153
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2-28-33, monoclonal

contains

15 mM sodium azide

species reactivity

Ciona intestinalis, rat, chicken, Caenorhabditis elegans, Strongylocentrotus purpuratus, bovine, human

enhanced validation

independent ( Antibodies)
Learn more about Antibody Enhanced Validation

technique(s)

indirect immunofluorescence: 1:1,000 using cultured chicken fibroblasts or cultured human foreskin fibroblasts, western blot: 1:100

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

β-Tubulin belongs to the subfamily of tubulin, which is the major building block of microtubules. β-tubulin has a molecular weight of 55kDa. β-Tubulin structure is characterized with core of two β-sheets enclosed by α-helices. It also contains an N-terminal domain with the guanine nucleotide-binding region, an intermediate domain with taxol-binding site, and a C-terminal domain that contains the binding surface for molecular motor proteins.Human β-tubulin consists of seven isoforms (βI (class I), βII (class II), βIII (class III), βIVa (class IVa), βIVb (class IVb), βV (class V), and βVI (class VI)).

Immunogen

sarkosyl-resistant ribbons from Strongylocentrotus purpuratus (sea urchin) sperm axonemes.

Application

Monoclonal Anti-β-Tubulin antibody produced in mouse has been used in western blotting and fluorescence immunocytochemical staining.

Biochem/physiol Actions

Mutation in the gene leads to various neuronal migration disorders such as lissencephaly, pachygyria and polymicrogyria malformations. B2702 peptide binds to β-tubulin and inhibits NK cell cytotoxicity and it influences microtubule polymerization, which damages cytoskeleton organization and chaperone-like activity of tubulin. β-1 tubulin also known as class VI or TUBB1, plays a vital role in platelet production, and is considered to be a potential biomarker candidate for immune thrombocytopenia (ITP).
The antibody recognizes the two major and one of the minor β‑tubulin isotypes of the free-living soil nematode C. elegans. It also binds to Sarkosyl-resistant ribbons from Strongylocentrotus purpuratus sperm axonemes, to neuronal axons of the larva of an ascidian (Ciona intestinalis) and to tubulin of chicken and mammals, cultured human fibroblasts, bovine and rat brain tissue. In C.elegans preparations, the antibody does not recognize tubulin that has been heat denatured in the presence of SDS and β-mercaptoethanol, though it is bound to tubulin separated on isoelectric focusing gels. It reacts with heat-denatured and reduced mammalian preparations in immunoblotting.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T4026T7816T5201
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

Ciona intestinalis, rat, chicken, Caenorhabditis elegans, Strongylocentrotus purpuratus, bovine, human

species reactivity

human, rat, frog, moth, mouse, plant, rabbit, chicken, bovine, wheat, sea urchin, hamster

species reactivity

Xenopus, chicken, human, hamster, canine, mouse, bovine, rat

species reactivity

human, rat, frog, moth, mouse, plant, rabbit, chicken, bovine, wheat, sea urchin, hamster

Gene Information

human ... TUBB1(81027), TUBB2A(7280), TUBB2C(10383)
rat ... Tubb2(29212), Tubb2c(296554)

Gene Information

human ... TUBB(203068), TUBB1(81027), TUBB1(81027), TUBB1(81027), TUBB1(81027), TUBB2A(7280), TUBB2A(7280), TUBB2A(7280), TUBB2A(7280), TUBB2C(10383), TUBB2C(10383), TUBB2C(10383), TUBB2C(10383)
mouse ... Tubb1(104068), Tubb1(104068), Tubb1(104068), Tubb1(104068), Tubb2a(22151), Tubb2a(22151), Tubb2a(22151), Tubb2a(22151), Tubb2c(227613), Tubb2c(227613), Tubb2c(227613), Tubb2c(227613)
rat ... Tubb2(29212), Tubb2(29212), Tubb2(29212), Tubb2(29212), Tubb2c(296554), Tubb2c(296554), Tubb2c(296554), Tubb2c(296554)

Gene Information

human ... TUBB(203068)
mouse ... Tubb4(22153)

Gene Information

human ... TUBB(203068), TUBB1(81027), TUBB1(81027), TUBB1(81027), TUBB1(81027), TUBB2A(7280), TUBB2A(7280), TUBB2A(7280), TUBB2A(7280), TUBB2C(10383), TUBB2C(10383), TUBB2C(10383), TUBB2C(10383)
mouse ... Tubb1(104068), Tubb1(104068), Tubb1(104068), Tubb1(104068), Tubb2a(22151), Tubb2a(22151), Tubb2a(22151), Tubb2a(22151), Tubb2c(227613), Tubb2c(227613), Tubb2c(227613), Tubb2c(227613), Tubb3(22152)
rat ... Tubb2(29212), Tubb2(29212), Tubb2(29212), Tubb2(29212), Tubb2c(296554), Tubb2c(296554), Tubb2c(296554), Tubb2c(296554)

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

purified immunoglobulin

clone

2-28-33, monoclonal

clone

TUB 2.1, monoclonal

clone

SAP.4G5, monoclonal

clone

TUB 2.1, monoclonal

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Birth of Piglets After Transfer of Embryos Cryopreserved by Cytoskeletal Stabilization and Vitrification
Dobrinsky JR
Biology of Reproduction, 62, 564-570 (2000)
The beta 1 tubulin R307H single nucleotide polymorphism is associated with treatment failures in immune thrombocytopenia (ITP).
Basciano PA
British Journal of Haematology, 160, 237-243 (2013)
Thomas Stahnke et al.
PloS one, 12(2), e0172592-e0172592 (2017-02-24)
In glaucoma surgery, fibrotic processes occur, leading to impairment of liquid outflow. Activated fibroblasts are responsible for postoperative scarring. The transforming growth factor-β (TGF-β) pathway plays a key role in fibroblast function, differentiation and proliferation. The aim of this study



Global Trade Item Number

SKUGTIN
T5293-.2ML04061838099921
T5293-100UL04061838026927

Questions

Reviews

No rating value

Active Filters