Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.32
UNSPSC Code:
41116158
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice
Pricing and availability is not currently available.
species reactivity
mouse
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
shipped in
wet ice
storage temp.
−20°C
General description
Immunoglobulin G (IgG) is the most prevalent protein found in the human serum. About 10–20% of plasma protein accounts for IgG.[1] IgG1 is the major Ig isotype in murine serum.[2] It resembles human IgG4 functionally. Murine immunoglobulin G1 (IgG1) is capable of blocking the complex formation of IgG2a, IgG2b, and IgG3 with C1q. C1q-mediated complement activation by IgG2a can be blocked by murine IgG1. Because of their likely inhibitory influence on both complement and classical Fcγ receptor (FcγR) activation, murine IgG1 may have the greatest inhibitory capacity toward all other IgG types.[3] The antibody pair provided in this kit recognizes mouse IgG1.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| species reactivity mouse | species reactivity mouse | species reactivity mouse | species reactivity human |
| assay range inter-assay cv: <10% | assay range inter-assay cv: <10% | assay range inter-assay cv: <10% | assay range inter-assay cv: <12% |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| RAB0802-1KT | 04061835165797 |




