Skip to Content
Merck

Skip To

P7367

PNGase F from Elizabethkingia meningoseptica

BioReagent, ≥95% (SDS-PAGE)

Synonym(s):

N-Glycosidase F, PNGase F from Chryseobacterium meningosepticum, PNGase F from Flavobacterium meningosepticum, Peptide N-glycosidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.32
EC Number:
MDL number:
Assay:
≥95% (SDS-PAGE)
Biological source:
bacterial (Elizabethkingia meningoseptica)
Concentration:
≥300 units/mL, ≥50 units/mL
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Elizabethkingia meningoseptica)

Quality Level

conjugate

(N-linked)

product line

BioReagent

assay

≥95% (SDS-PAGE)

form

powder

shelf life

≥1 weeks at 2‑8 °C (for a reconstituted solution >500 units/ml), ≥1 yr at 2‑8 °C, Solution is stable for at least 3 freeze-thaw cycles

mol wt

~36 kDa

concentration

≥300 units/mL, ≥50 units/mL

optimum pH

~8.6

shipped in

wet ice

storage temp.

2-8°C

General description

Proteomics Grade PNGase F has been extensively purified and lyophilized from dilute potassium phosphate buffer to produce a stable product. The product is free from glycerol and other stabilizers, and contains very low levels of buffer salts. This highly purified material is excellent for N-linked deglycosylation of glycoproteins or glycopeptides in gel, in solution, or on blot membranes.

Application

PNGase F from Elizabethkingia meningoseptica has been used:
  • for de-N-glycosylation of Zika20virus E protein[1]
  • to evaluate coxsackievirus and adenovirus receptor glycosylation using CAR-expressing COS cells[2]
  • to verify the N-linked glycosylation of MHC class 1 polypeptide-related sequence A (MICA)[3]

Suitable for both proteomics and glycobiology use; compatible with MALDI-TOF MS analysis
Used to deglycosylate protein.

Biochem/physiol Actions

Cleaves an entire glycan from a glycoprotein provided the glycosylated asparagine moiety is substituted on its amino and carboxyl terminus with a polypeptide chain.

Other Notes

One unit will catalyze the release of N-linked oligosaccharides from 1 nanomole of denatured ribonuclease B in one minute at 37°C at pH 7.5 monitored by SDS-PAGE. One Sigma unit of PNGase F activity is equal to 1 IUB milliunit.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G5166G1549F8435
biological source

bacterial (Elizabethkingia meningoseptica)

biological source

bacterial (Elizabethkingia miricola)

biological source

-

biological source

-

assay

≥95% (SDS-PAGE)

assay

-

assay

-

assay

-

form

powder

form

buffered aqueous solution

form

ready-to-use solution

form

lyophilized powder

concentration

≥300 units/mL, ≥50 units/mL

concentration

-

concentration

-

concentration

-

optimum pH

~8.6

optimum pH

-

optimum pH

-

optimum pH

-

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Terry Nguyen-Khuong et al.
Glycoconjugate journal, 35(6), 499-509 (2018-11-24)
Analysis of glycans via a porous graphitized carbon liquid chromatography (PGC-LC) coupled with electrospray ionization (tandem) mass spectrometry (ESI-MS(/MS)) is a powerful analytical method in the field of glycomics. Isobaric glycan structures can be identified reliably with the help of
Lars Andresen et al.
Journal of immunology (Baltimore, Md. : 1950), 188(4), 1847-1855 (2012-01-10)
NKG2D ligand surface expression is important for immune recognition of stressed and neotransformed cells. In this study, we show that surface expression of MICA/B and other NKG2D ligands is dependent on N-linked glycosylation. The inhibitor of glycolysis and N-linked glycosylation
Katherine J D Ashbourne Excoffon et al.
Journal of virology, 81(11), 5573-5578 (2007-03-23)
The coxsackievirus and adenovirus receptor (CAR) is both a viral receptor and homophilic adhesion protein. The extracellular portion of CAR consists of two immunoglobulin (Ig)-like domains, each with a consensus sequence for N-glycosylation. We used chemical, genetic, and biochemical studies



Global Trade Item Number

SKUGTIN
P7367-300UN04061834394679
P7367-10UN04061832020181
P7367-50UN04061834394686

Questions

Reviews

No rating value

Active Filters