Skip to Content
Merck

Skip To

LIG1

DNA Ligation Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.53
UNSPSC Code:
12352204
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

usage

1 kit sufficient for 150 ligation reactions

shipped in

wet ice

storage temp.

−20°C

General description

Sigma′s DNA Ligation Kit contains all of the reagents necessary to perform DNA ligation reactions. Sticky-end ligations are more efficient than blunt-end ligations; these can be facilitated with the addition of PEG.

Application

Suitable for:
  • Joining fragments of DNA into a cloning vector
  • Mutagenesis
  • Gene anyalysis and structure-function relationships

Biochem/physiol Actions

One of the most important steps in the cloning process is the ligation of linear DNA into a cloning vector. DNA ligations are performed by incubating DNA fragments with appropriately linearized cloning vectors in the presence of buffer, ATP, and DNA ligase. Many parameters affect ligations such as the relative ratio of insert to vector, the quality and type of the DNA ends, the temperature of ligation and the concentration of DNA.

Other Notes

Sufficient for 150 reactions:
  • 300uL 10X Ligation Buffer (D2176) in 250 mM Tris-HCl (pH 7.8) 100 mM MgCl2, and 10 mM dithiothreitol
  • 3 x 100 units T4 DNA Ligase (D2886) in 50% glycerol with 10 mM Tris-HCl (pH 7.5) 50 mM KCl, and 1 mM dithiothreitol
  • 3 x 100 uL 10 mM ATP ( A3702)
  • 50 uL Control pBR322 DNA, HAE III Digest (D9430) 0.5 ug/ul in 10 mM Tris-HCl (pH 8.0), and 1 mM EDTA
  • 1.5 mL 24% (w/v) PEG Solution, (P 2454)
  • 1.5 mL Molecular Biology Grade Water (W4502)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
LIG211635379001RDLIG-RO
usage

1 kit sufficient for 150 ligation reactions

usage

 kit sufficient for 50 ligation reactions

usage

sufficient for 40 reactions (DNA ligation)

usage

sufficient for 160 reactions (04898125001), sufficient for 40 reactions (04898117001)

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

dry ice

shipped in

-

shipped in

-

grade

Molecular Biology

grade

Molecular Biology

grade

-

grade

-

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

-


pictograms

Health hazard

signalword

Danger

hcodes

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
APREST82550-100UL04061835842766
HPA041431-100UL04061836326272
HPA048071-25UL04061841229773
SAB2501284-100UG04061835791569
AV54307-100UL04061836205683
HPA041431-25UL04061841563747
HPA048071-100UL04061831727210
WH0003978M1-100UG04061831634136
LIG1-1KT04061833967218
EHU153921-20UG04061831366884
EMU072751-20UG04061828482016
EMU072751-50UG04061828256723
EHU153921-50UG04061828409938