Skip to Content
Merck

Skip To

F4375

Ficoll® PM 400

Type 400

Synonym(s):

Polysucrose 400

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

non-animal source

type

Type 400

form

powder, spray-dried

optical activity

[α]20/D 56.5°, c = 1% (w/v) in water

technique(s)

tissue culture: suitable

color

white to off-white

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

suitability

suitable for component for culture media

storage temp.

room temp

General description

Ficoll 400 is a highly branched polymer formed by the copolymerization of sucrose and epichlorohydrin. Ficoll 400 is completely non-ionic. Because of the abundance of hydroxyl groups, Ficoll 400 is very hydrophilic and extremely water-soluble. The most common application for Ficoll 400 is as a density gradient medium for the separation and isolation of eukaryotic cells, organelles and bacterial cells. Density ranges up to 1.2 g/ml can be attained. It has also been utilized in a variety of other applications.

Application

Ficoll® PM 400 is used together with sodium diatrizoate (Paque) to create Ficoll-Paque a density mixture used for the separation of cells and organelles, especially blood derived lymphocytes and monocytes. It may be used in tissue engineering and macromolecular conformation research for the development, evaluation and use of macromolecular crowding (MMC) systems and configurations. Ficoll PM-400 may be used in cell freeze-drying research.
Used for cell separation and organelle isolation.

Features and Benefits

Because of the high molecular weight and low content of dialyzable material, Ficoll has a much lower permeability towards cell membranes than sucrose. Therefore, cells can be expected to collect at a lower density in Ficoll gradients than in sucrose gradients. Because of its low membrane permeability and low osmotic pressure, separations in Ficoll normally result in better preservation of cell function and morphology.

Preparation Note

Concentrations of 50% (w/v) can be attained in water. Ficoll should be added slowly with constant stirring.
Sigma tests the solubility of Ficoll 400 at 1 g in 10 ml of deionized water yielding a clear to slightly hazy, colorless to faint yellow solution. Ficoll is stable in alkaline and neutral solutions. At pH values below 3, it is rapidly hydrolyzed, particularly at elevated temperatures. Ficoll can be sterilized by autoclaving at a neutral pH, at 110oC for 30 minutes. Strong oxidizing and reducing agents are to be avoided.

Other Notes

A nonionic synthetic polymer of sucrose.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Ficoll is a registered trademark of Cytiva

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F1418F8636341691
technique(s)

tissue culture: suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

technique(s)

-

biological source

non-animal source

biological source

-

biological source

-

biological source

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

solubility

water: soluble

solubility

-

solubility

water: soluble

suitability

suitable for component for culture media

suitability

-

suitability

-

suitability

-

form

powder, spray-dried

form

lyophilized powder

form

lyophilized powder

form

solid


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters