Skip to Content
Merck

Skip To

F1283

Fetal Bovine Serum

Fetal Bovine Serum, USA origin, Ultra Low IgG

Synonym(s):

FBS, FCS, Fetal Calf Serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Biological source:
bovine fetus
Origin:
USA origin
Sterility:
sterile-filtered
Shipped in:
dry ice
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine fetus

Quality Level

sterility

sterile-filtered

product line

FBS Select

composition

Bovine IgG, <5 μg/mL , Hemoglobin, ≤20 mg/dL

origin

USA origin

storage condition

avoid repeated freeze/thaw cycles, protect from light

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

pH

6.8-8.1

osmolality

260-330 mOsm/kg H2O

shipped in

dry ice

storage temp.

−20°C

General description

Animal serum is commonly used to supplement basal media formulations for the optimal growth of many cell types in vitro.

Fetal Bovine Serum (FBS) is the most common serum used to supplement cell culture media due to its high nutritional content. Although it is relatively low in protein, FBS is effective in promoting and sustaining growth of vertebrate mammalian and insect cells.

IgG is the most common immunoglobulin present in FBS. Elevated levels of IgG may cause problems within certain recombinant protein or monoclonal antibody purification processes. For applications where IgG may interfere or cause issues, we have depleted IgG levels in this fetal bovine serum by sequential chromatography down to a level of < 5 ug/mL, while maintaining performance equivalent to regular FBS.

Packaging

Packaged in sterilized, graduated plastic bottles and sealed with a tamper indicator. Bottles are immediately frozen and stored at -10 to -40 C.

Preparation Note

The fetal blood is aseptically collected and allowed to clot under controlled conditions. After centrifugation, the raw (unfiltered) serum is decanted from the clot, pooled and immediately frozen.

Frozen raw serum is thawed under controlled conditions and then processed through a series of membrane filters in descending pore size. FBS is filtered through three 0.1 μm filters before packaging.

Analysis Note

Endotoxin and hemoglobin tested

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F244212103CF4135
origin

USA origin

origin

USA origin

origin

USA origin

origin

USA origin

biological source

bovine fetus

biological source

bovine fetus

biological source

bovine fetus

biological source

bovine fetus

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | hybridoma: suitable, cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | hybridoma: suitable, cell culture | insect: suitable, cell culture | mammalian: suitable

sterility

sterile-filtered

sterility

sterile-filtered

sterility

-

sterility

sterile-filtered

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice


Still not finding the right product?

Explore all of our products under Fetal Bovine Serum


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content





Global Trade Item Number

SKUGTIN
F1283-500ML04061838095213
F1283-100ML04061838069009

Questions

Reviews

No rating value

Active Filters