Skip to Content
Merck
All Photos(1)

Key Documents

F1141

Sigma-Aldrich

Bovine Fibronectin

from bovine plasma, liquid, suitable for cell culture

Synonym(s):

Fibronectin

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42020141
NACRES:
NA.75

Product Name

Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture

biological source

bovine plasma

Quality Level

sterility

sterile-filtered

product line

BioReagent

form

solution

packaging

pkg of 1 mg
pkg of 2 mg
pkg of 5 mg

technique(s)

cell culture | mammalian: suitable

surface coverage

1‑5 μg/cm2

UniProt accession no.

Binding Specificity

Peptide Source: Collagen

Peptide Source: Fibrinogen

shipped in

wet ice

storage temp.

2-8°C

Gene Information

bovine ... FN1(280794)

Looking for similar products? Visit Product Comparison Guide

General description

Fibronectin is a ubiquitous protein of the extracellular matrix. It is a mosaic protein, that is made up of three types of repeating modular consensus amino acid sequences. Fibronectins belongs to a class of high-molecular-weight glycoproteins. Cellular fibronectin is expressed by fibroblasts and plasma fibronectin is usually liberated by hepatocytes.

Application

Fibronectin (FN) bovine plasma has been used in:
  • fibronectin and adsorption analysis (ELISA (enzyme-linked immunosorbent assay))
  • transwell migration
  • fibronectin assay

Biochem/physiol Actions

Fibronectins participate in cell adhesion, cytoskeletal organization, oncogenic transformation, cell migration, phagocytosis, hemostasis, and embryonic differentiation.

Analysis Note

Protein homogeneity is evaluated by immunoelectrophoresis.

Other Notes

Product F1141, Fibronectin from bovine plasma is a solution in 0.05 M TBS pH 7.5.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of subnano-, nano-and submicron-surface features on osteoblast differentiation of bone marrow mesenchymal stem cells
Khang D, et al.
Biomaterials, 33(26), 5997-6007 (2012)
Enhanced fibronectin adsorption on carbon nanotube/poly (carbonate) urethane: independent role of surface nano-roughness and associated surface energy
Khang D, et al.
Biomaterials, 28(32), 4756-4768 (2007)
CCL2/CCR2 chemokine signaling coordinates survival and motility of breast cancer cells through Smad3 and p42/44MAPK dependent mechanisms
Fang W B, et al.
The Journal of biological chemistry, jbc-M112 (2012)
FIBRONECTIN AUTOANTIBODIES
Autoantibodies, 221-251 (2007)
Tests to predict preeclampsia
Chesley's Hypertensive Disorders in Pregnancy, 221-251 (2015)

Articles

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

Fibronectin (FN) plays crucial roles in extracellular matrix fibril assembly and cellular interactions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service