Skip to Content
Merck

Skip To

F0503

5-Fluoro-2′-deoxyuridine

thymidylate synthase inhibitor

Synonym(s):

floxuridine, 2′-Deoxy-5-fluorouridine, FUDR, Floxuridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C9H11FN2O5
CAS Number:
Molecular Weight:
246.19
NACRES:
NA.77
PubChem Substance ID:
EC Number:
200-072-5
Beilstein/REAXYS Number:
90221
MDL number:
UNSPSC Code:
12352208
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥99% (HPLC)

form

powder

mp

148 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(F)C(=O)NC2=O

InChI

1S/C9H11FN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1

InChI key

ODKNJVUHOIMIIZ-RRKCRQDMSA-N

Gene Information

human ... TYMS(7298)
mouse ... Tyms(22171)

General description

5-Fluoro-2′-deoxyuridine, also called floxuridine elicits DNA-directed cytotoxicity in cancer cells. Floxuridine is effective for treating liver cancer and eliminating virulence of Staphylococcus aureus. Dipeptide prodrugs combination of floxuridine with gemcitabine are more cell permeable and display enhanced anti-proliferative activity. Floxuridine is effective on solid tumours and advanced stage cancers.

Application

5-Fluoro-2′-deoxyuridine has been used as a mitotic inhibitor in schwann cell proliferation, glia proliferation and nonneuronal cells in dorsal root ganglion cultures.

Biochem/physiol Actions

Antineoplastic drug that acts as a potent inhibitor of thymidylate synthetase Resistance to FUdR can develop in cancer cell cultures, among other means, by low-level Mycoplasma infection.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR25893433331271008
biological source

synthetic (organic)

biological source

-

biological source

-

biological source

-

assay

≥99% (HPLC)

assay

-

assay

≥98% (HPLC)

assay

-

Gene Information

human ... TYMS(7298)
mouse ... Tyms(22171)

Gene Information

-

Gene Information

-

Gene Information

human ... TYMS(7298)

Quality Level

200

Quality Level

300

Quality Level

100

Quality Level

-

form

powder

form

powder

form

solid

form

-

storage temp.

room temp

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

-


Still not finding the right product?

Explore all of our products under 5-Fluoro-2′-deoxyuridine


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
F0503-250MG04061833611234
F0503-1G04061833611227
F0503-100MG04061833611203

Questions

1–2 of 2 Questions  
  1. Once I reconstitute this product in water, can I store it long term?

    1 answer
    1. A sterile 2% aqueous solution of product is reported to be stable for up to 2 weeks at 2–8 °C, protected from light. Long-term stability at -20 °C is not established for this product, although an external source suggests potential stability for up to 6 months at -20 °C. This information has not been validated.
      Please see the link below to review additional information available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/716/505/f0503pis.pdf

      Helpful?

  2. Is it possible to sterile filter (0.2 mikrometer) this compound for safer use in cell culture?

    1 answer
    1. This product is soluble in water at 50 mg/mL. Solutions may be filter sterilized and are stable for 2 weeks at 2-8 °C. Solutions should be protected from light.
      See the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/716/505/f0503pis.pdf

      Helpful?

Reviews

No rating value

Active Filters