Skip to Content
Merck

Skip To

E8145

Ethylene glycol-bis(β-aminoethyl ether)-N,N,N′,N′-tetraacetic acid tetrasodium salt

≥97%

Synonym(s):

EGTA tetrasodium, Egtazic acid tetrasodium, Ethylene-bis(oxyethylenenitrilo)tetraacetic acid tetrasodium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H20N2O10Na4
CAS Number:
Molecular Weight:
468.28
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥97%

technique(s)

RNA purification: suitable

solubility

H2O: soluble 10%, clear, colorless

application(s)

life science and biopharma

SMILES string

[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCOCCOCCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C14H24N2O10.4Na/c17-11(18)7-15(8-12(19)20)1-3-25-5-6-26-4-2-16(9-13(21)22)10-14(23)24;;;;/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24);;;;/q;4*+1/p-4

InChI key

BAOGCDPNNOBWQZ-UHFFFAOYSA-J

Application

EGTA is suitable for use:
  • in media for chelating Ca2+ to facilitate efficient incorporation of Sr2+ into the oocytes[1]
  • as a component of phosphatase wash buffer used to wash magnetic beads during dephosphorylation and radiolabeling of RNA segments cross-linked to immunoprecipitated proteins[2]
  • as a component of oocyte activation media
A chelating agent useful for the determination of calcium in the presence of magnesium.
A chelating agent useful for the determination of calcium in the presence of magnesium. Repeated washing with EGTA-buffer can experimentally deplete cultured cells of Ca2+.

Biochem/physiol Actions

Ethylene glycol-bis(β-aminoethyl ether)-N,N,N,N-tetra acetic acid tetrasodium salt (EGTA) is a calcium chelator that binds to Ca2+ in the presence of Mg2+ at a ratio of 1:1.
Ethylene glycol-bis(β-aminoethyl ether)-N,N,N′,N′-tetra acetic acid tetrasodium salt (EGTA) dissociates mucin complex and is useful in immunohistochemical studies. It suppresses root growth and mitotic index in germinating wheat seeds. It is used as a decalifying agent in root canal biomechanical preparation and is effective compared to EDTA and cyclohexanediaminetetraacetic acid (CDTA).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E4378E0396ED4S
technique(s)

RNA purification: suitable

technique(s)

-

technique(s)

-

technique(s)

-

assay

≥97%

assay

≥97.0%

assay

≥97 .0%

assay

≥95%

Quality Level

200

Quality Level

400

Quality Level

400

Quality Level

200

application(s)

life science and biopharma

application(s)

-

application(s)

-

application(s)

-

solubility

H2O: soluble 10%, clear, colorless

solubility

1 M NaOH: 1.1 g/10 mL, clear, colorless

solubility

1 M NaOH: 0.1 M, clear, colorless, H2O: soluble (increases as pH is increased)

solubility

soluble, clear, colorless (32g plus 200ml of water)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet


Sayaka Wakayama et al.
Methods in enzymology, 476, 151-169 (2010-08-10)
Somatic cell nuclear transfer (SCNT) has become a unique and powerful tool for epigenetic reprogramming research and gene manipulation in animals. Although the success rates of somatic cloning have been inefficient and the mechanism of reprogramming is still largely unknown
Shirley P and Saunders T L
Advanced Protocols For Animal Transgenesis, 271-277 (2011)
Schmid, R.W. et al.
Analytical Chemistry, 29, 264-264 (1957)



Global Trade Item Number

SKUGTIN
E8145-50G04061833608654
E8145-10G04061833608647

Questions

Reviews

No rating value

Active Filters