Skip to Content
Merck

Skip To

D9891

Doxycycline hyclate

Synonym(s):

6-Desoxy-5-hydroxytetracycline hydrochloride hemihydrate hemiethanolate, Doxycycline hydrochloride hemiethanolate hemihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
CAS Number:
Molecular Weight:
512.94
UNSPSC Code:
51284027
NACRES:
NA.85
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5702728
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Level

assay

≥93.5% (HPLC), 95.0-102.0% anhydrous basis (ethanol free based)

form

powder

storage condition

(Tightly closed. Dry. )

color

yellow to yellow-green

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, mycoplasma, parasites

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].[H]O[H].CCO.C[C@@H]1C2[C@H](O)C3[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]3(O)C(O)=C2C(=O)c4c(O)cccc14.C[C@@H]5C6[C@H](O)C7[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]7(O)C(O)=C6C(=O)c8c(O)cccc58

InChI

1S/2C22H24N2O8.C2H6O.2ClH.H2O/c2*1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;1-2-3;;;/h2*4-7,10,14-15,17,25,27-29,32H,1-3H3,(H2,23,31);3H,2H2,1H3;2*1H;1H2/t2*7-,10?,14?,15-,17-,22-;;;;/m00..../s1

InChI key

HALQELOKLVRWRI-ZVACAFRPSA-N

General description

Doxycycline Hyclate, a synthetic tetracycline antibiotic, demonstrates broad-spectrum antimicrobial activity against various bacteria by binding to the 30S and potentially the 50S ribosomal subunits. This binding disrupts the aminoacyl-tRNA binding to the mRNA-ribosome complex, effectively inhibiting bacterial protein synthesis, thereby halting growth and replication.

Beyond its antimicrobial effects, Doxycycline Hyclate serves as an inhibitor of matrix metalloproteinases (MMPs), enzymes crucial for collagen breakdown in connective tissues. Studies on corneal erosion and rheumatoid arthritis showcase its significant reduction in MMP-9 and MMP-8 levels, respectively. This MMP-inhibiting property helps preserve collagen integrity, preventing excessive tissue degradation and supporting tissue repair.

In cell biology, Doxycycline Hyclate impacts smooth muscle cells, enhancing adhesion and influencing the reorganization of fibrillar collagen matrices. Its influence on apicoplast gene expression in Plasmodium falciparum, the malaria-causing parasite, adds a dimension of significance in infectious disease research. Additionally, the inhibition of tissue formation in rat studies contributes to its relevance in understanding tissue development.

Moreover, Doxycycline Hyclate holds potential in metabolomics research, considering its multi-faceted effects on various cellular processes. In the broader context of biochemical research, its versatile actions make it a valuable tool for investigating molecular mechanisms and signaling pathways.

Application

Doxycycline hyclate has been used for the induction of glioma cells and human embryonic kidney 293T cells (HEK293T).
Doxycycline hyclate is a synthetic oxytetracycline derivative. It has been used to eliminate Borrelia burgdorferi and Anaplasma phagocytophilum in rodent reservoirs and to eliminate Ixodes scapularis ticks[1]. It is a broad spectrum inhibitor used to inhibit matrix metalloproteinases[2] (MMP), such as type 1 collagenase in studies on wound healing and tissue remodeling.

Biochem/physiol Actions

Mode of Action: Doxycycline hyclate inhibits the inflammatory response to the Lyme Disease Spirochete Borrelia burgdorferi.

Activity Spectrum: Effective against a broad spectrum inhibitor of matrix metalloproteinases in vivo.

Features and Benefits

  • High-quality antibiotic suitable for multiple research applications
  • Ideal for Cell Biology, Metabolomics, and Biochemical research.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1145D52071226003
antibiotic activity spectrum

Gram-negative bacteria, mycoplasma, Gram-positive bacteria, parasites

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

-

mode of action

protein synthesis | interferes

mode of action

-

mode of action

-

mode of action

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

-

form

powder

form

-

form

powder

form

-

assay

≥93.5% (HPLC), 95.0-102.0% anhydrous basis (ethanol free based)

assay

-

assay

-

assay

-

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Doxycycline hyclate


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Regulation of glioma cell invasion by 3q26 gene products PIK 3 CA, SOX 2 and OPA 1
Schaefer T, et al.
Brain Pathology (2018)
Kranthi Marella Panth et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 116(3), 462-466 (2015-07-15)
The central hypothesis of "radiomics" is that imaging features reflect tumor phenotype and genotype. Until now only correlative studies have been performed. The main objective of our study is to determine whether a causal relationship exists between genetic changes and
Basheer Khadaroo et al.
Nature cell biology, 11(8), 980-987 (2009-07-15)
The ends of linear eukaryotic chromosomes are protected by telomeres, which serve to ensure proper chromosome replication and to prevent spurious recombination at chromosome ends. In this study, we show by single cell analysis that in the absence of telomerase



Global Trade Item Number

SKUGTIN
D9891-100G04061833591291
D9891-1G04061835544332
D9891-5G04061833591314
D9891-25G04061833591307
D9891-10G04061835544325

Questions

1–10 of 14 Questions  
  1. What grade is D9891? Is it food-grade?

    1 answer
    1. This product is not food-grade. Unless otherwise stated, this product is intended for research use only. It is not suitable for any other purpose, which includes, but is not limited to, unauthorized commercial use, in vitro diagnostic use, or therapeutic use in humans or animals.
      Unfortunately, a food grade Doxycycline hyclate product is not currently offered.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. What is the potency in µg/mg of Product D9891, Doxycycline Hyclate?

    1 answer
    1. The potency of this product has not been tested.

      Helpful?

  4. Does this powder if stored in dark at 4 degree expires? Doxycycline hyclate, Product D9891

    1 answer
    1. This product is assigned an expiration date of 4 years from the date of Quality Release. The expiration date is lot specific and reported on the product label and the Certificate of Analysis. The recommended storage temperature range is 2 - 8°C. Please see the link below to review additional information available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/162/885/d9891pis.pdf

      Helpful?

  5. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  6. What is the expiration date of Doxycycline hyclate(powder, not solution) from the date of purchase?

    1 answer
    1. This product is not assigned an expiration date. It is assigned a recommended retest date of approximately 3.5 years after the quality release date. A recommended retest date is the period of time during which the product is expected to remain within established stability specifications, provided that it has been stored under defined conditions. After the Retest Date, product samples should be examined to ensure that the product is still in compliance with the established specifications. For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

  7. Once reconstituted in water, can Doxycycline hyclate be stored long-term at -20°C ?

    1 answer
    1. Aqueous solutions of Doxycycline are light sensitive and should be used within 48 hours. The long term stability of aqueous solutions at -20°C has not been evaluated. However, the Doxycycline HCl Ready Made solution is prepared at 100 mg/ml in DMSO. This product is assigned a shelf life of 1 year and is stored at -20°C. See the links below to review the product datasheet for D9891, as well as a sample Certificate of Analysis for the Ready Made Solution, D3072.

      Datasheet, D9891
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/162/885/d9891pis.pdf

      Sample Certificate, D3072
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/D3072-1ML0000229139.pdf

      Helpful?

  8. What is the solubility of Doxycycline hyclate, Product D9891?

    1 answer
    1. Doxycycline hyclate is soluble in water at up to 50 mg/ml.

      Helpful?

  9. Is doxycycline hyclate soluble in DMSO?

    1 answer
    1. According to literature sources, doxycycline hyclate is soluble in DMSO at a concentration of 100 mM.

      Helpful?

  10. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

1–10 of 14 Questions  

Reviews

No rating value

Active Filters