Skip to Content
Merck

Skip To

D9503

3,4-Dihydroxy-DL-phenylalanine

Synonym(s):

3-(3,4-Dihydroxyphenyl)-DL-alanine, DL-3-Hydroxytyrosine, DL-DOPA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(HO)2C6H3CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
197.19
NACRES:
NA.32
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-566-0
MDL number:
Beilstein/REAXYS Number:
3204800
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Level

mp

270-272 °C (lit.), 290 °C (dec.) (lit.)

SMILES string

NC(Cc1ccc(O)c(O)c1)C(O)=O

InChI

1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)

InChI key

WTDRDQBEARUVNC-UHFFFAOYSA-N

Biochem/physiol Actions

3,4-dihydroxyphenylalanine is an immediate precursor of dopamine, which is a product of tyrosine hydroxylase.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H8653T6062SAB2900445
form

powder

form

-

form

powder

form

buffered aqueous solution

Gene Information

human ... DRD1(1812), DRD2(1813), DRD3(1814), DRD4(1815), DRD5(1816)
mouse ... DRD1(13488), DRD2(13489), DRD3(13490), DRD4(13491), DRD5(13492)
rat ... DRD1(24316), DRD2(24318), DRD3(29238), DRD4(25432), DRD5(25195)

Gene Information

human ... DRD3(1814)

Gene Information

human ... DRD2(1813), DRD3(1814), DRD4(1815), HTR2A(3356), HTR2C(3358)

Gene Information

human ... DRD5(1816)

mp

270-272 °C (lit.), 290 °C (dec.) (lit.)

mp

-

mp

243 °C (dec.) (lit.)

mp

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Steven L Saville et al.
Nanoscale, 5(5), 2152-2163 (2013-02-08)
It has been recently reported that for some suspensions of magnetic nanoparticles the transverse proton relaxation rate, R(2), is dependent on the time that the sample is exposed to an applied magnetic field. This time dependence has been linked to
Luis García-Fernández et al.
Advanced materials (Deerfield Beach, Fla.), 25(4), 529-533 (2012-11-10)
Inspired by the amino acid 2-chloro-4,5-dihydroxyphenylalanine (Cl-DOPA), present in the composition of the proteinaceous glue of the sandcastle worm Phragmatopoma californica, a simple strategy is presented to confer antifouling properties to polymer surfaces using (but not releasing) a bioinspired biocide.
Dopamine formation, from its immediate precursor 3, 4-dihydroxyphenylalanine, along the rat digestive tract
Vieira-Coelho MA and Soares-da-Silva P
Fundamental & Clinical Pharmacology, 7(5), 235-243 (1993)



Global Trade Item Number

SKUGTIN
D9503-1G04061826738467
D9503-25G04061833590775
D9503-5G04061833590782

Questions

Reviews

No rating value

Active Filters