Skip to Content
Merck

Skip To

D4645

Sodium 3,5-dichloro-2-hydroxybenzenesulfonate

≥98% purity (TLC), powder

Synonym(s):

3,5-Dichloro-2-hydroxybenzenesulfonic acid, DCHBS, DHBS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H3Cl2NaO4S
CAS Number:
Molecular Weight:
265.05
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
EC Number:
259-416-8
MDL number:
Beilstein/REAXYS Number:
3640643
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sodium 3,5-dichloro-2-hydroxybenzenesulfonate, used for peroxide measurement

Quality Segment

assay

≥98% (TLC)

form

powder

technique(s)

titration: suitable

color

white to off-white

pH

7 (20 °C, 53 g/L)

mp

>300 °C

solubility

H2O: 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Oc1c(Cl)cc(Cl)cc1S([O-])(=O)=O

InChI

1S/C6H4Cl2O4S.Na/c7-3-1-4(8)6(9)5(2-3)13(10,11)12;/h1-2,9H,(H,10,11,12);/q;+1/p-1

InChI key

NMWCVZCSJHJYFW-UHFFFAOYSA-M

General description

Sodium 3,5-dichloro-2-hydroxybenzenesulfonate is used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of uric acid in serum and urine samples.

Application

Used in conjunction with 4-aminoantipyrine (4-AAP) and hydrogen peroxide (H2O2) for chromogenic quantitation of peroxidase in coupled enzymatic reactions. Component of Trinder reagent for use with peroxidase to measure generation of hydrogen peroxide in automated systems.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B1125S4506S5752
technique(s)

titration: suitable

technique(s)

titration: 90% using HCLO4 (Dry Basis)

technique(s)

titration: suitable

technique(s)

-

color

white to off-white

color

white to beige

color

white to off-white

color

-

solubility

H2O: 50 mg/mL

solubility

H2O: 50 mg/mL

solubility

H2O: 350 mg/mL, clear to slightly hazy

solubility

water: 1 mg/mL, blue

form

powder

form

powder

form

powder

form

powder

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

2-8°C


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters