Skip to Content
Merck

Skip To

D4540

Dimethyl sulfoxide

≥99.5% (GC), suitable for plant cell culture

Dimethyl sulfoxide
1 of 2 reviewers received a sample product or took part in a promotion

Synonym(s):

DMSO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
UNSPSC Code:
12191502
NACRES:
NA.71
PubChem Substance ID:
EC Number:
200-664-3
Beilstein/REAXYS Number:
506008
MDL number:
Assay:
≥99.5% (GC)
Technique(s):
cell culture | plant: suitable, cryopreservation: suitable
Bp:
189 °C (lit.)
Vapor pressure:
0.42 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

assay

≥99.5% (GC)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

technique(s)

cell culture | plant: suitable, cryopreservation: suitable

impurities

<0.1% water (Karl Fischer)

color

colorless

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

solubility

H2O: miscible (completely)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Application

Dimethyl sulfoxide has been used:
  • in the preparation of bisphenol A (BPA)
  • in cell proliferation assay (MTT assay)
  • in chemosensitivity assay
  • as the vehicle control in all experiments like apoptosis assay
  • in ethoxyresorufin-O-deethylation (EROD) assay

Biochem/physiol Actions

DMSO (dimethyl sulfoxide), an organic solvent possesses several pharmaceutical activities such as analgesic, diuretic, bacteriostatic, membrane-penetrant, anti-inflammatory, vasodilatory and cholinesterase inhibitory effects. It is a by-product of the lumber industry and used as an industrial solvent. DMSO also serves as a therapeutic agent to treat interstitial cystitis and arthritis.

Disclaimer

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D26530193441640-M
Quality Level

200

Quality Level

300

Quality Level

100

Quality Level

-

assay

≥99.5% (GC)

assay

≥99.7%

assay

≥99.9% (GC), ≥99.99995% (metals basis)

assay

≥99.9% (GC)

technique(s)

cell culture | plant: suitable, cryopreservation: suitable

technique(s)

cell culture | hybridoma: suitable, cryopreservation: suitable

technique(s)

-

technique(s)

GC/GC: suitable

solubility

H2O: miscible (completely)

solubility

H2O: miscible (completely)

solubility

H2O: miscible (completely)

solubility

-

form

liquid

form

liquid

form

liquid

form

liquid

density

1.10 g/mL (lit.)

density

1.10 g/mL (lit.)

density

1.099-1.101 g/mL at 20 °C, 1.10 g/mL (lit.)

density

1.10 g/mL (lit.)


Still not finding the right product?

Explore all of our products under Dimethyl sulfoxide


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Pharmacotherapy of joint and tendon disease
Equine Sports Medicine and Surgery E-Book: Basic and clinical sciences of the equine athlete, 473-473 (2013)
Curcumin promotes apoptosis, increases chemosensitivity, and inhibits nuclear factor kappaB in esophageal adenocarcinoma
Hartojo W, et al.
Translational Oncology, 3(2), 99-108 (2010)
Evaluation of the sub-lethal toxicity of Cu, Pb, bisphenol A and polychlorinated biphenyl to the marine dinoflagellate Cochlodinium polykrikoides
Ebenezer V and Ki J S
Algae, 27(1), 63-63 (2012)



Global Trade Item Number

SKUGTIN
D4540-1L04061838259295
D4540-100ML04061838259288
D4540-500ML04061838259301

Questions

1–10 of 11 Questions  
  1. Can I use this product (suitable for plant cell culture) for cyropreservation of mammalian cells? Does cryopreservation of mammalian cells require sterile-filtered DMSO?

    1 answer
    1. D4540 is listed as suitable for cryopreservation.
      However, D2650 is already pre-sterile and has documentation with a protocol for cryopreservation of human and animal cell lines.

      See these links:
      https://www.sigmaaldrich.com/product/sigma/d2650
      https://www.sigmaaldrich.com/product/sigma/d2650#product-documentation

      Look for the Product Information Sheet. Also, see this link for more information:
      https://www.sigmaaldrich.com/products/cell-culture-and-analysis/cell-culture-supplements-and-reagents/cell-freezing-reagents

      Helpful?

  2. Can I autoclave this product?

    1 answer
    1. DMSO is not autoclavable in neat form. It is highly unlikely that microorganisms would survive in this environment. Should sterilization be necessary a PTFE, Polypropylene, or Nylon filter membrane are recommended for sterile-filtration.

      Helpful?

  3. What is the molarity of DMSO?

    1 answer
    1. DMSO will have a molarity of 14.1M, based on a density of 1.1 g/mL and a molecular weight of 78.13 g/mol.

      Helpful?

  4. Does DMSO have any synthetic utility?

    1 answer
    1. DMSO has been used to oxidize thiols and disulfides to sulfonic acids, hydrolysis of epoxides, thioalkylation of phenols, and oxidation of primary alcohols and halides, and esters of primary alcohols to aldehydes.

      Helpful?

  5. What is the DMSO dissolved in?

    1 answer
    1. DMSO is not dissolved in any solvent. It is a pure chemical compound that is a liquid at room temperature.

      Helpful?

  6. Where does DMSO come from?

    1 answer
    1. DMSO is known to have been isolated from tree bark in the past. It is now a commericially synthesized solvent.

      Helpful?

  7. Dimethyl sulfoxide (DMSO) should be a liquid.  Why did it arrive as a solid?

    1 answer
    1. DMSO supercools easily, and re-melts slowly at room temperature. The melting point is 16-19 °C. The solidified product can be re-liquefied by warming to room temperature without detriment to the product.

      Helpful?

  8. What type of filter is used to sterile-filter Dimethyl sulfoxide (DMSO)?

    1 answer
    1. DMSO can be sterile-filtered through a 0.2 micron PTFE filter. The sterility testing is per the current US Pharmacopoeia method.

      Helpful?

  9. What concentration of DMSO do I use in a cell freezing media?

    1 answer
    1. DMSO is usually used at 1-10%, depending on the cell line.  For the cell freezing media that we offer, it is used at 8.7% concentration.

      Helpful?

  10. Is Dimethyl sulfoxide a sterile product?

    1 answer
    1. No, this product is not considered sterile.  Product Nos. D2560 and D2438 are sterile-filtered.

      Helpful?

1–10 of 11 Questions  

Reviews

Active Filters

  1. San Diego
    • Review 1
    • Votes 4
    5 out of 5 stars.

    Works on mammalian cells

    Good for mammalian cell culture treatments of compound

    Helpful?

  2. 1 Ratings-only review