Skip to Content
Merck

Skip To

D3695

DMOG

≥98% (HPLC), powder, HIF-hydroxylase inhibitor

Synonym(s):

Dimethyloxalylglycine, N-(Methoxyoxoacetyl)-glycine methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H9NO5
CAS Number:
Molecular Weight:
175.14
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

DMOG, ≥98% (HPLC)

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: >30 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

COC(=O)CNC(=O)C(=O)OC

InChI

1S/C6H9NO5/c1-11-4(8)3-7-5(9)6(10)12-2/h3H2,1-2H3,(H,7,9)

InChI key

BNJOZDZCRHCODO-UHFFFAOYSA-N

Application

DMOG has been used:
  • in hypoxia-inducible factor (HIF) activity assay
  • to examine its effects on the degradation of HIF-1α and renal regeneration[1]
  • in DMOG preconditioning of adipose tissues
  • as vehicle control for the primary liquid culture of CD34+ cells
  • for endothelial cell stimulation

Dimethyloxalylglycine (DMOG) has been used as an inhibitor of ten-eleven translocation 3 (TET3) protein.

Biochem/physiol Actions

DMOG is a cell permeable prolyl-4-hydroxylase inhibitor, which upregulates HIF (hypoxia-inducible factor).
DMOG is a cell permeable prolyl-4-hydroxylase inhibitor, which upregulates HIF (hypoxia-inducible factor). The protein level of HIF-1α subunit is post-transcriptionally regulated by prolyl and asparaginyl hydroxylase (PAH). Suppression of PAH activity increases endogenous HIF-1α levels. DMOG is a cell permeable, competitive inhibitor of prolyl hydroxylase domain-containing proteins (PHDs and HIF-PHs). It has been discovered that the DMOG posseses neuroprotective effect on NFG deprived cell cultures through preservation of glucose metabolism. DMOG also attenuates myocardial injury in a rabbit ischemia reperfusion model. DMOG is more potent than the older inhibitor 4-Phenyl-pyridine-2,5-dicarboxylic acid (R395889; Sigma-Aldrich rare chemicals library). The IC50 is 5.18 μM.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G0673P0064SML1306
form

powder

form

powder

form

solid

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

H2O: >30 mg/mL

solubility

DMSO: >20 mg/mL

solubility

DMSO: >10 mg/mL

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

-


Still not finding the right product?

Explore all of our products under DMOG


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Metabolic reprogramming by HIF-1 activation enhances survivability of human adipose-derived stem cells in ischaemic microenvironments
Chen C, et al.
Cell Proliferation, 50(5), e12363-e12363 (2017)
Perparim Limani et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 22(23), 5887-5897 (2016-11-03)
Tumor hypoxia activates hypoxia-inducible factors (Hifs), which induce a range of malignant changes including vascular abnormalities. Here, we determine whether inhibition of the hypoxic tumor response through myo-inositol trispyrophosphate (ITPP), a compound with antihypoxic properties, is able to cause prolonged
Jianping Peng et al.
PloS one, 12(5), e0178147-e0178147 (2017-05-26)
Acute kidney injury (AKI) leads to a worse prognosis in diabetic patients compared with prognoses in non-diabetic patients, but whether and how diabetes affects kidney repair after AKI remains unknown. Here, we used scratch-wound healing and transwell migration models to



Global Trade Item Number

SKUGTIN
D3695-50MG04061833563526
D3695-10MG04061833563519

Questions

1–2 of 2 Questions  
  1. Is it suitable to use D3695 to induce hypoxia?

    1 answer
    1. Upon verification, D3695 is known to induce hypoxia. For further reference, you can refer the following citation: "Biomed Pharmacother. 2020 Jan;121:109606. doi: 10.1016/j.biopha.2019.109606. Epub 2019 Nov 25. Prodrug of epigallocatechin-3-gallate alleviates choroidal neovascularization via down-regulating HIF-1α/VEGF/VEGFR2 pathway and M1 type macrophage/microglia polarization."

      Helpful?

  2. Bonjour, Quelle est la durée de conservation à -20°C de ce réactif ? Merci

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment.

      For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters