Skip to Content
Merck

Skip To

CELLMM2

FLAG® M Purification Kit

For Mammalian expression systems.

Synonym(s):

Anti-ddddk, Anti-dykddddk

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

technique(s)

protein extraction: suitable

shipped in

wet ice

storage temp.

−20°C

Application

The FLAG Purification Kit utilizes CelLytic Reagents for rapid and efficient cell lysis and protein extraction and ANTI-FLAG® M2 affinity gel for affinity purification of active FLAG fusion proteins.

Learn more product details in our FLAG® application portal.

Features and Benefits

  • Includes ready to use reagents, columns, and a detailed protocol for affinity purification of FLAG fusion proteins.
  • ANTI-FLAG M2 Affinity Gel allows efficient binding of FLAG fusion proteins without the need for preliminary steps and calibrations.
  • Two alternatives for efficient elution conditions (by acidic conditions or by competition with FLAG peptide).

Packaging

Sufficient for 3-5 uses of 1-ml affinity purification column.

Other Notes

Kit contains CelLytic™ M for cell lysis and protein extraction from Mammalian cells.

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
CelLytic is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
TP0010FLAGIPT1E6155
technique(s)

protein extraction: suitable

technique(s)

-

technique(s)

immunoprecipitation (IP): suitable

technique(s)

-

shipped in

wet ice

shipped in

-

shipped in

wet ice

shipped in

dry ice

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization.
    SDS

  • Purified 3xFLAG® peptide, ≥95% (HPLC), lyophilized powder
    SDS

  • ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
    SDS

  • Chromatography columns, general-purpose, volume 10 mL, Overall H 5 in.
    SDS

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Erik G Puffenberger et al.
PloS one, 7(1), e28936-e28936 (2012-01-27)
The Clinic for Special Children (CSC) has integrated biochemical and molecular methods into a rural pediatric practice serving Old Order Amish and Mennonite (Plain) children. Among the Plain people, we have used single nucleotide polymorphism (SNP) microarrays to genetically map
Amitava Mukherjee et al.
PLoS pathogens, 7(3), e1001311-e1001311 (2011-03-26)
The host innate immune response to viral infections often involves the activation of parallel pattern recognition receptor (PRR) pathways that converge on the induction of type I interferons (IFNs). Several viruses have evolved sophisticated mechanisms to attenuate antiviral host signaling
Severa Bunda et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(36), E3785-E3794 (2014-08-27)
Mutations in Ras GTPase and various other components of the Ras signaling pathways are among the most common genetic alterations in human cancers and also have been identified in several familial developmental syndromes. Over the past few decades it has



Global Trade Item Number

SKUGTIN
CELLMM2-1KT04061833531679

Questions

  1. What is the approximate yield of protein (in ug) if you start with a one 10 cm dish?

    1 answer
    1. The actual yield of purified FLAG-tagged protein from a 10 cm dish using the CELLMM2 kit will depend on the expression level of the protein in the cells. The kit’s resin can bind up to 600–1000 µg per 1 mL, but typical yields from mammalian cells are often in the 100–500 µg range per dish. For precise yield, measuring the protein concentration in the lysate before and after purification is recommended.

      Helpful?

Reviews

No rating value

Active Filters