Skip to Content
Merck

Skip To

C9750

β-Carotene

synthetic, ≥93% (UV), powder

Synonym(s):

β,β-Carotene, Provitamin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
UNSPSC Code:
12352205
NACRES:
NA.79
PubChem Substance ID:
EC Number:
230-636-6
Beilstein/REAXYS Number:
1917416
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Level

assay

≥93% (UV)

form

powder

potency

~1,600,000 units vitamin A per g

color

dark red to brown

mp

178-179 °C

solubility

hexane: 1.1 mg/mL, DMSO: 30 μg/mL

storage temp.

−20°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C=C/C2=C(C)CCCC2(C)C)C(C)(C)CCC1

InChI

1S/C40H56/c1-31(19-13-21-33(3)25-27-37-35(5)23-15-29-39(37,7)8)17-11-12-18-32(2)20-14-22-34(4)26-28-38-36(6)24-16-30-40(38,9)10/h11-14,17-22,25-28H,15-16,23-24,29-30H2,1-10H3/b12-11+,19-13+,20-14+,27-25+,28-26+,31-17+,32-18+,33-21+,34-22+

InChI key

OENHQHLEOONYIE-JLTXGRSLSA-N

General description

Carotenoids are an important class of naturally occurring pigments found in fruits and vegetables. Among the carotenoids, β-carotene has the greatest provitamin A potential and functions to reduce the risk of cancer, cardiovascular diseases and macular degeneration. Low water solubility and poor chemical stability attributes to low absorption of β-carotene from food. β-carotene along with digestible lipids enriches its bio accessibility. Emulsions or nano emulsions encapsulates effectively delivers lipophilic β-carotene and increases its bioavailability.[1] β-carotene has anti-oxidant potential and reduces noise induced hearing loss and progression of macular degeneration.

Application

β-carotene has been used:
  • to generate nano emulsions to evaluate their bioavailability potential[1]
  • as a standard, for β-carotene evaluation in blood and skin samples using high performance liquid chromatography (HPLC),
  • as a dietary supplement of vitamin-A in young rats by HPLC
  • as a dietary supplement in mice to reduce hearing loss and hair cell damage

Biochem/physiol Actions

The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro. However, in vivo it appears to act either as an antioxidant or a prooxidant depending on cellular environment. It reduces the incidence of many cancers, but enhances lung cancer incidence in smokers.
The most important of the provitamins A.

Packaging

Packaged in serum vials.

Disclaimer

Purchase minimum quantity required. Discard one month after opening.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1239217538V-058
biological source

synthetic

biological source

-

biological source

-

biological source

-

assay

≥93% (UV)

assay

-

assay

≥97% (HPLC)

assay

-

form

powder

form

solid

form

solid

form

liquid

solubility

hexane: 1.1 mg/mL, DMSO: 30 μg/mL

solubility

-

solubility

DMSO: soluble, cyclohexane: soluble

solubility

-

mp

178-179 °C

mp

-

mp

-

mp

-

color

dark red to brown

color

-

color

dark red

color

-


Still not finding the right product?

Explore all of our products under β-Carotene


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

217.9 °F - closed cup

flash_point_c

103.3 °C - closed cup

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Colleen G Le Prell et al.
Free radical biology & medicine, 42(9), 1454-1463 (2007-03-31)
Free radical formation in the cochlea plays a key role in the development of noise-induced hearing loss (NIHL). The amount, distribution, and time course of free radical formation have been defined, including a clinically significant formation of both reactive oxygen
Qingqing Liu et al.
eLife, 5 (2016-05-25)
Gap junctions are widely distributed in the brains across species and play essential roles in neural information processing. However, the role of gap junctions in insect cognition remains poorly understood. Using a flight simulator paradigm and genetic tools, we found
Nutrient-enhanced diet reduces noise-induced damage to the inner ear and hearing loss
Le Prell CG, et al.
Translational Research, 158(1), 38-53 (2011)



Global Trade Item Number

SKUGTIN
C9750-10G04061835543748
C9750-5G04061835543755

Questions

1–4 of 4 Questions  
  1. Can this be used to prepare standard curves for the determination of β-carotene content in foods?

    1 answer
    1. The minimum purity of this product is 93% by UV-Vis Spectrophotometry. This item is intended for use in general laboratory applications and has not been tested for suitability as a standard. Suitability would have to be determined by the end user. Note that product V-058, a 100 ug/mL Certified Reference Material, is tested as a standard in HPLC/UV, LC/MS, and GC/FID headspace and may be of interest. Please see the link below to review this product option:
      https://www.sigmaaldrich.com/US/en/product/cerillian/v058

      Helpful?

  2. Is this food grade material?

    1 answer
    1. This product is not a food grade material. A food grade β-Carotene product is not currently offered.

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  4. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters