Skip to Content
Merck

Skip To

C8392

Clove oil

Synonym(s):

Eugenia spp., Oil of cloves

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

oil

Quality Segment

refractive index

n20/D 1.532 (lit.)

bp

251 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

lipid type

oils

Application


  • Clove Essential Oil: Chemical Profile, Biological Activities, Encapsulation Strategies, and Food Applications: This review examines the chemical characteristics of clove essential oil, its biological effects, encapsulation methods, and its application in food industry, emphasizing its potential as a natural preservative and flavor enhancer (Liñán-Atero et al., 2024).

  • Chitosan-Oxidized Pullulan Hydrogels Loaded with Essential Clove Oil: Synthesis, Characterization, Antioxidant and Antimicrobial Properties: This study describes the creation of chitosan-based hydrogels incorporating clove oil, detailing their synthesis, characterization, and their effective antioxidant and antimicrobial properties, useful in biomedical applications and food preservation (Suflet et al., 2024).

  • Sustained release, antimicrobial, and antioxidant properties of modified porous starch-based biodegradable polylactic acid/polybutylene adipate-co-terephthalate/thermoplastic starch active packaging film: Research on biodegradable packaging films enhanced with clove oil, highlighting its sustained release capabilities and benefits in enhancing antimicrobial and antioxidant properties to extend the shelf life of packaged foods (Gui et al., 2024).

  • Corn cob nanocellulose packaging for increasing the shelf life of food products: This article explores the use of corn cob nanocellulose as a packaging material, where clove oil could be utilized as a natural preservative to enhance the product′s shelf life and safety (Cheran et al., 2024).

Biochem/physiol Actions

Essential oil used for its medicinal properties. Immunomodulator and anti-inflammatory.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W23230024245377430
form

oil

form

-

form

liquid

form

liquid

density

1.04 g/mL at 25 °C (lit.)

density

1.05 g/mL at 25 °C

density

0.86 g/mL at 25 °C (lit.)

density

1.013 g/mL at 25 °C (lit.)

Quality Level

100

Quality Level

300, 400

Quality Level

100

Quality Level

100

refractive index

n20/D 1.532 (lit.)

refractive index

n20/D 1.532 (lit.)

refractive index

n20/D 1.4670-1.4780

refractive index

n20/D 1.529 (lit.)

bp

251 °C (lit.)

bp

251 °C (lit.)

bp

153-175 °C (lit.)

bp

150 °C (lit.)

lipid type

oils

lipid type

-

lipid type

-

lipid type

-


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

239.0 °F - closed cup

flash_point_c

115 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1B



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
C8392-500ML04061833522240
C8392-100ML04061838255020

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is it 100% Clove oil? what are % content of all isomeres?

    1 answer
    1. The product is considered to be pure clove oil, but the isomer and impurity content are not characterized. Please see the sample SDS and specification sheet to see what properties are tested.

      Helpful?

  4. Can you provide a certified information of % content of all isomeres (eugenol, iso-eugenol and methyl-eugenol) in the Clove Oil C8392 ? Thanks !

    1 answer
    1. All of the characterization that is performed is listed on the COA, an example of which can be found on the product page in the 'Documentation' section. The individual components are not quantified in this case.

      Helpful?

Reviews

No rating value

Active Filters