Skip to Content
Merck

Skip To

C8176

Collagenase from Clostridium histolyticum

Sigma Blend Type L, ≤1.0 FALGPA units/mg solid

Synonym(s):

Clostridiopeptidase A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-582-9
MDL number:
EC Number:
Specific activity:
≤1.0 FALGPA units/mg solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

powder

caseinase activity

≥40 units/mg solid

specific activity

≤1.0 FALGPA units/mg solid

mol wt

68-130 kDa

storage temp.

−20°C

Application

Collagenase from Clostridium histolyticum, or Clostridiopeptidase A, has been used in a study to assess the synthesis and in vitro evaluation of macromolecular antitumour derivatives based on phenylenediamine mustard. Clostridiopeptidase A has also been used in a study to investigate the enzymatic debridement of burn wounds with collagenase.

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.
Effective release of cells from tissue requires the action of collagenase enzymes and the neutral protease. Collagenase is activated by four gram atom calcium (Ca2+) per mole enzyme. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa. The pH optimum is 6.3-8.8. The enzyme is typically used to digest the connective components in tissue samples to liberate individual cells. Ethylene glycol-bis(β-aminoethyl ether)-N,N,N′,N′-tetraacetic acid (EGTA)4; β-mercaptoethanol; glutathione, reduced; thioglycolic acid, sodium; and 2,2′-dipyridyl; 8-hydroxyquinoline are known to inhibit the enzyme activity.

Other Notes

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7926C8051C2014
specific activity

≤1.0 FALGPA units/mg solid

specific activity

≥2.0 FALGPA units/mg solid

specific activity

≥1.0 FALGPA units/mg solid

specific activity

≥1 FALGPA units/mg solid, ≥125 CDU/mg solid

form

powder

form

powder

form

powder

form

lyophilized powder

mol wt

68-130 kDa

mol wt

68-130 kDa

mol wt

68-130 kDa

mol wt

68-125 kDa

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

caseinase activity

≥40 units/mg solid

caseinase activity

≤10 units/mg solid

caseinase activity

≥10 units/mg solid

caseinase activity

-


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Coşkun Ozcan et al.
Burns : journal of the International Society for Burn Injuries, 28(8), 791-794 (2002-12-05)
Seventy-eight pediatric burn patients treated by enzymatic debridement with collagenase clostridiopeptidase A (CCA), were compared to 41 patients those burn wounds were excised surgically. Patients whose burn wounds were initially assessed as partial-thickness at admission were enrolled in the study.
N I Solov'eva et al.
Bioorganicheskaia khimiia, 20(3), 303-309 (1994-03-01)
Interactions of collagenases I and II (clostridiopeptidases) from Clostridium histolyticum with hexapeptide substrates in which some L-proline residues are replaced by their D-analogues, as well as with the tripeptide chloromethyl ketone Z-Gly-Pro-Gly-CH2Cl were studied. A role of stereochemistry of the
Ruth M Williams et al.
STAR protocols, 2(2), 100414-100414 (2021-04-20)
In order to process samples by fluorescence-activated cell sorting (FACS), it is essential to obtain a single-cell suspension of dissociated cells. Numerous protocols and commercial reagents are available; however, each requires optimization for specific tissue types. Here, we describe an



Global Trade Item Number

SKUGTIN
C8176-1G04061826682203
C8176-500MG04061832700304
C8176-100MG04061832700281
C8176-25MG04061832700298
C8176-5G04061831157635

Questions

Reviews

No rating value

Active Filters