Skip to Content
Merck

Skip To

C5679

Casein from bovine milk

BioReagent, suitable for insect cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.75
EC Number:
232-555-1
MDL number:
Form:
powder
Biological source:
bovine milk
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine milk

Quality Level

product line

BioReagent

form

powder

technique(s)

cell culture | insect: suitable

mp

280 °C (dec.) (lit.)

UniProt accession no.

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

General description

Casein is a calcium phosphate-binding phosphoprotein present in mammalian milk. It constitutes about 80% of the total bovine milk. Bovine milk is made of four types of casein such as, αs1-, αs2-, β-, and κ-caseins.

Application

Casein from bovine milk has been used as a component of normal medium for Drosophila culture , in the preparation of αS1-casein (CAS1_Bovin) , as a source of nitrogen (N) in experiment diets .

Biochem/physiol Actions

Numerous experimental applications including use as a blocking agent in immunochemistry, recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
Partial gastrointestinal digestion of casein is a rich source of bioactive peptides, such as β-casomorphin. However, bovine casein is not homogeneous; variants A1 and B do lead to production of β-casomorphin 7 production, while A2 does not.

Analysis Note

essentially vitamin free

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C4765C6905C0406
technique(s)

cell culture | insect: suitable

technique(s)

activity assay: suitable, electrophoresis: suitable, immunocytochemistry: suitable

technique(s)

activity assay: suitable

technique(s)

isoelectric focusing (IEF): suitable

biological source

bovine milk

biological source

bovine milk

biological source

bovine milk

biological source

bovine milk

form

powder

form

liquid

form

essentially salt-free, lyophilized powder

form

lyophilized powder

Gene Information

bovine ... CSN1S1(282208), CSN1S2(282209), CSN2(281099), CSN3(281728)

Gene Information

bovine ... CSN1S1(282208), CSN1S2(282209), CSN2(281099), CSN3(281728)

Gene Information

bovine ... CSN2(281099), CSN3(281728)

Gene Information

cow ... MKCAS(100531419), MKCASP(105755569)

UniProt accession no.

P02662, P02666, P02663, P02668

UniProt accession no.

P02662, P02663, P02666, P02668

UniProt accession no.

P02666, P02668

UniProt accession no.

-

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200


Still not finding the right product?

Explore all of our products under Casein from bovine milk


Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Potential of casein as a carrier for biologically active agents
Glkab T K and Boratynski J
Topics in Current Chemistry, 375(4), 71-71 (2017)
Casein and Peptides Derived from Casein as Antileukaemic Agents
Ledesma M E, et al.
Journal of Oncology, 2019(4), 71-71 (2019)
S Elsayed et al.
Molecular immunology, 41(12), 1225-1234 (2004-10-16)
T cell recognition patterns of CAS1_Bovin, its limited hydrolysis, oxidized, reduced/alkylated, cyanogen bromide cleavage fractions and synthetic peptides were examined. Thirteen overlapping peptides covering the intact molecule, with chain lengths varied between 17 and 20 AA, were prepared by f-moc



Global Trade Item Number

SKUGTIN
C5679-500G04061833502433

Questions

Reviews

No rating value

Active Filters