Skip to Content
Merck

Skip To

B9428

Monoclonal Anti-Bin1 antibody produced in mouse

clone 99D, ascites fluid

Synonym(s):

Anti-Amphiphysin II, Anti-SH3P9, Anti-ampl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
99D, monoclonal
Application:
ELISA (i), FACS, ICC, IHC (f), IP, WB
Citations:
10
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

99D, monoclonal

mol wt

antigen 65-75 kDa

contains

15 mM sodium azide

species reactivity

rabbit, mouse, rat, avian, human

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable using nucleoplasm, subnuclear punctate compartment, and cytoplasm, immunohistochemistry (frozen sections): suitable, immunoprecipitation (IP): suitable, indirect ELISA: suitable, western blot: 1:50,000 using a rat brain cytosol preparation

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BIN1(274)
mouse ... Bin1(30948)
rat ... Bin1(117028)

General description

Monoclonal Anti-Bin1 (SH3P9, Amphiphysin II, amphl) (mouse IgG2b isotype) is derived from the 99D hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a recombinant polypeptide, containing amino acids 189-398 of human Bin1. Bin1 [Box-dependent myc-interacting protein-1, also known as SH3P9, amphiphysin II and amphl (amphiphysin-like)] is a novel protein that has features of a tumor suppressor. The human BIN1 gene has been mapped to chromosome 2q14. In human and rodent cells it appears as a short-lived (half-life of approx. 2 hrs) monomeric phosphoprotein (approx. 70 kDa). Bin1 molecule appears in different isoforms or conformations, and the localization varies between normal and tumor cells. In normal cells, Bin1 is predominantly nucleoplasmic, but is also present in a subnuclear compartment. In a panel of tumor cells that express Bin1, the predominant localization of the molecule is in the subnuclear compartment.

Immunogen

recombinant polypeptide containing amino acids 189-398 of human Bin1.

Application

Monoclonal Anti-Bin1 antibody produced in mouse has been used in:
  • western blotting
  • enzyme linked immunosorbent assay (ELISA)
  • immunoprecipitation
  • immunocytochemistry
  • immunohistochemistry
  • flow cytometry

Biochem/physiol Actions

Box-dependent myc-interacting protein-1 (Bin1) interacts with and inhibits the oncogenic activity of the myc oncoprotein that has a major role in many human cancers. The loss of Bin1 may contribute to growth deregulation in cancer cells in carcinoma of the breast, colon, lung, cervix, prostate and liver. In bin1 locus mid-2q region that is deleted in approx. 42% of metastatic prostate cancers, and at the syntenic murine locus, in >90% of radiation-induced myeloid leukemias. In addition to the role of Bin1 in controlling neoplastic cell growth and cell cycle by the inhibition of transformation by both myc and the adenovirus E1A protein. Deficits in expression are functionally significant, because ectopic Bin1 can inhibit the growth of tumor cells that lack endogenous expression. Bin1 has also been found to be highly expressed in murine skeletal muscle; overexpression of Bin1 promotes myotube formation and upregulation of myosin heavy chain, while interference with Bin1 expression significantly impairs these processes.
The antibody reacts specifically with a 33-residue segment of the myc-binding domain (MBD, amino acids 323-356) of the human Bin1 molecule. It does not react with the Bin1-related 45 kDa polypeptide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M4308SAB1408547HPA005437
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

Gene Information

human ... BIN1(274)
mouse ... Bin1(30948)
rat ... Bin1(117028)

Gene Information

human ... MDM2(4193)
mouse ... Mdm2(17246)
rat ... Mdm2(246362)

Gene Information

human ... BIN1(274)

Gene Information

human ... BIN1(274)

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

rabbit

species reactivity

rabbit, mouse, rat, avian, human

species reactivity

human, rat, mouse (weak)

species reactivity

human

species reactivity

human

clone

99D, monoclonal

clone

SMP14, monoclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Loss of heterozygosity and tumor suppressor activity of Bin1 in prostate carcinoma
Ge K, et al.
International Journal of Cancer. Journal International Du Cancer, 86(2), 155-161 (2000)
Accumulation of caveolin-3 protein is limited in damaged muscle in chicken muscular dystrophy
Matsumoto H, et al.
Comparative Biochemistry and Physiology. Part A, Molecular & Integrative Physiology, 157(1), 68-72 (2010)
Characterization of bridging integrator 1 (BIN1) as a potential tumor suppressor and prognostic marker in hepatocellular carcinoma
Pan K, et al.
Molecular Medicine, 18(3), 507-518 (2012)



Global Trade Item Number

SKUGTIN
B9428-.2ML04061837864650

Questions

Reviews

No rating value

Active Filters