Skip to Content
Merck

Skip To

B6429

Casein Blocking Buffer 10x

for Northern and Southern blotting, solution

Synonym(s):

10x Blocking Buffer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.54
UNSPSC Code:
12161700
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Level

form

solution

technique(s)

Southern blotting: suitable

storage temp.

room temp

General description

Casein is a phosphoprotein found in milk. It is classified into four subtypes, including αS1-, αS2-, β-, and κ-caseins.[1]
Sigma’s 10x Blocking Buffer is a casein-based blocking agent routinely used to reduce non-specific binding of detection reagents[1] to the Northern and Southern Blot membranes.

Application

Casein Blocking Buffer 10x has been used in:

  • western blotting[2][3][4]
  • enzyme linked immunosorbent assay (ELISA)
  • immunohistochemistry

Features and Benefits

  • Better blocking performance than dried milk or BSA
  • Easy-to-use concentrate
  • Compatible with neutral and positively-charged membranes
  • Compatible with chemiluminescent and colorimetric detection assays

Preparation Note

Dilute 10x Blocking Buffer with molecular biology grade water before use.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7594G766393017
technique(s)

Southern blotting: suitable

technique(s)

Southern blotting: suitable

technique(s)

-

technique(s)

Southern blotting: suitable

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

solution

form

powder blend

form

powder blend

form

solution

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Choline availability modulates human neuroblastoma cell proliferation and alters the methylation of the promoter region of the cyclin-dependent kinase inhibitor 3 gene
Niculescu MD, et al.
Journal of Neurochemistry, 89(5), 1252-1259 (2004)
Tanja Kalic et al.
The journal of allergy and clinical immunology. In practice, 8(9), 3084-3092 (2020-05-12)
Fish collagen is widely used in medicine, cosmetics, and the food industry. However, its clinical relevance as an allergen is not fully appreciated. This is likely due to collagen insolubility in neutral aqueous solutions, leading to low abundance in commercially
Mihai D Niculescu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 20(1), 43-49 (2006-01-06)
The availability of choline during critical periods of fetal development alters hippocampal development and affects memory function throughout life. Choline deficiency during fetal development reduces proliferation and migration of neuronal precursor cells in the mouse fetal hippocampus and these changes



Global Trade Item Number

SKUGTIN
B6429-500ML04061833434796

Questions

1–8 of 8 Questions  
  1. Is Casein Blocking Buffer (B6429) compatible with SSC-based pre-hybridization or hybridization buffers for Northern blotting (e.g. 6× SSC, 0.5% SDS) with directly fluorescently labeled probes, or is it intended only for post-hybridization blocking steps?

    1 answer
    1. The 10X Casein Blocking Buffer is recommended for both colorimetric and chemiluminescent detection. It is used as a blocking solution for detection, which occurs after hybridization and is suitable for Northern and Southern applications.

      Please also review our Technical Document, Northern & Southern Blot Protocols, available at the link below:
      https://www.sigmaaldrich.com/technical-documents/protocol/protein-biology/gel-electrophoresis/southern-and-northern-blotting

      Helpful?

  2. Hi, how much time can be the casein blocking buffer 10X stored from its opening?

    1 answer
    1. This product does not have an expiration date once opened. The datasheet indicates the following regarding storage and stability:

      Storage/Stability: Store the 10X Casein Blocking Buffer at room temperature. It is also stable when stored at 2–8°C. Casein may precipitate over time, which is normal and should not impact the performance of the 10X Blocking Buffer. If precipitation is observed, resuspension of the solids can be achieved by inverting the bottle several times or by vortexing for a few seconds.

      Helpful?

  3. Hi, would it be possible to know the % of casein in your Casein Blocking Buffer 10X Cat# B6429? Thank you.

    1 answer
    1. The composition of this product is proprietary and cannot be disclosed. However, it can be stated that the product is a blocking reagent derived from bovine milk casein and does not contain any serum albumin.

      Helpful?

  4. whats the percentage casein in 10X of B6429-500ml ?

    1 answer
    1. Unfortunately, the concentration of casein is considered proprietary.

      Helpful?

  5. Will my RNA samples and probes be safe with this buffer if we are using horseradish peroxidase and chemiluminescent detection?

    1 answer
    1. The 10X Casein Blocking Buffer is recommended for both colorimetric and chemiluminescent detection. It is used as a blocking solution for detection, which occurs after hybridization and is suitable for Northern and Southern applications.

      Helpful?

  6. What is the pH? What is the counter ion? Is there saline salt in the preparation (0.15M)?

    1 answer
    1. Unfortunately, the composition of this product is proprietary and can't be disclosed.

      Helpful?

  7. What is the % w/v casein in this solution?

    1 answer
    1. The casein concentration is 1.8% of the total solution.

      Helpful?

  8. What is the solution solvent? What grade of reagent is this in terms of sterility and associated testing?

    1 answer
    1. As mentioned in the product detail page, this product is a molecular biology-grade product. A solution is prepared by diluting the 10x Blocking Buffer with molecular biology-grade water.

      Helpful?

Reviews

No rating value

Active Filters