Skip to Content
Merck

Skip To

A9256

L-Aspartic acid

≥98% (HPLC)

L-Aspartic acid
1 of 1 reviewers received a sample product or took part in a promotion

Synonym(s):

(S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HO2CCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
133.10
NACRES:
NA.26
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-291-6
MDL number:
Beilstein/REAXYS Number:
1723530
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Aspartic acid, reagent grade, ≥98% (HPLC)

grade

reagent grade

Quality Segment

assay

≥98% (HPLC)

form

powder, crystals or chunks

color

white to off-white

mp

>300 °C (dec.) (lit.)

solubility

H2O: 5 mg/mL, 0.5 M HCl: 50 mg/mL (with heat), 1 M NaOH: 50 mg/mL

SMILES string

N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1

InChI key

CKLJMWTZIZZHCS-REOHCLBHSA-N

Gene Information

human ... CA1(759), CA2(760)
rat ... Grin2a(24409)

Application

L-aspartic acid has been used as one of the components of synthetic drop out media for culturing budding yeast. It has also been used to study non-enzymatic gluconeogenesis.

Biochem/physiol Actions

Principal neurotransmitter for fast synaptic excitation.
Aspartic acid is a non-essential amino acid. It is involved in the Urea cycle, Krebs cycle and in DNA metabolism.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7219A894951572
grade

reagent grade

grade

-

grade

-

grade

certified reference material, TraceCERT®

form

powder, crystals or chunks

form

powder

form

powder

form

-

assay

≥98% (HPLC)

assay

98.5-101.0%

assay

≥99% (HPLC)

assay

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

300

color

white to off-white

color

white to off-white

color

white to off-white

color

-

mp

>300 °C (dec.) (lit.)

mp

>300 °C (dec.) (lit.)

mp

>300 °C (dec.) (lit.)

mp

>300 °C (dec.) (lit.)


Still not finding the right product?

Explore all of our products under L-Aspartic acid


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. East Lansing MI
    • Reviews 4
    • Votes 5
    5 out of 5 stars.

    L-Aspartic acid

    L-Aspartic acid from sigma have high quality , presentation and easy to use

    Helpful?