Skip to Content
Merck
All Photos(4)

Key Documents

A8094

Sigma-Aldrich

L-Arginine

98.5-101.0%, suitable for cell culture, BioXtra, non-animal source

Synonym(s):

(S)-2-Amino-5-guanidinopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
174.20
Beilstein:
1725413
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%

biological source

non-animal source

Quality Level

Agency

USP/NF
meets EP testing specifications
meets USP testing specifications

product line

BioXtra

Assay

98.5-101.0%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

222 °C (dec.) (lit.)

solubility

H2O: 100 mg/mL

application(s)

agriculture
peptide synthesis
pharmaceutical (small molecule)

SMILES string

N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1

InChI key

ODKSFYDXXFIFQN-BYPYZUCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

L-Arginine has been used:
  • as a Roswell park memorial institute medium (RPMI) media component in the isolation and culture of peripheral blood mononuclear cells (PBMCs)
  • as a RPMI media component for tissue culture
  • in DMEM medium for the identification and quantification of phosphorylation sites by stable isotope labeling by amino acids in cell culture (SILAC) and LCMS/MS

Biochem/physiol Actions

L-Arginine is a conditionally essential amino acid, that play a vital role in the production of nitric oxide. It acts as a precursor for the synthesis of polyamines, agmatine and urea. L-Arginine is usually essential in children and non-essential in adults.
Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell-specific Labeling Enzymes for Analysis of Cell?Cell Communication in Continuous Co-culture
Tape CJ, et.al.
Molecular and Cellular Proteomics, 13(7), 1866-1876 (2014)
Anti-aging effects of L-arginine
Gad M Z
Journal of Advanced Research, 1(3), 169-177 (2010)
Requirement of the mTOR kinase for the regulation of Maf1 phosphorylation and control of RNA polymerase III-dependent transcription in cancer cells
Shor B, et al.
The Journal of biological chemistry, jbc-M109 (2010)
Requirement of the mTOR Kinase for the Regulation of Maf1 Phosphorylation and Control of RNA Polymerase III-dependent Transcription in Cancer Cells
Boris Shor
The Journal of Biological Chemistry, 285(20), 15380-15392 (2010)
Association of reactive oxygen and nitrogen intermediate and complement levels with apoptosis of peripheral blood mononuclear cells in lupus patients
Oates J C, et al.
Arthritis and Rheumatism, 56(11), 3738-3747 (2007)

Protocols

XTT assay protocol for measuring cell viability, proliferation, activation and cytotoxicity. Instructions for XTT reagent preparation and examples of applications.

MTT assay protocol for measuring cell viability, proliferation and cytotoxicity. Instructions for MTT reagent preparation and examples of applications.

MTT assay protocol for measuring cell viability, proliferation and cytotoxicity. Instructions for MTT reagent preparation and examples of applications.

MTT assay protocol for measuring cell viability, proliferation and cytotoxicity. Instructions for MTT reagent preparation and examples of applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service