Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352302
NACRES:
NA.25
Pricing and availability is not currently available.
Product Name
Sodium cacodylate Buffer pH 6.5; 50 mM – Isopropanol 15% solution,
form
liquid
Quality Level
final pH
6.5±0.2 ( in neat)
density
0.98 g/mL at 20 °C
storage temp.
2-8°C
Application
Sodium cacodylate Buffer pH 6.5; 50 mM − Isopropanol 15% solution has been used:
- in washing the overnight fixed pancreas sections for transmission electron microscope assay
- in the preparation of fixation and post-fixation buffers and washing of cells for scanning electron microscopy (SEM)
- in diluting glutaraldehyde for the fixation of bacterial biofilm for SEM analysis
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
98.6 °F - closed cup
flash_point_c
37.0 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Emodin attenuated severe acute pancreatitis via the P2X ligand-gated ion channel 7/NOD-like receptor protein 3 signaling pathway
Zhang Q, et al.
Oncology Reports, 41(1), 270-278 (2019)
Surgical retrieval, isolation and in vitro expansion of human anterior cruciate ligament-derived cells for tissue engineering applications
Gupta A, et al.
Journal of Visualized Experiments, e51597-e51597 (2014)
Oral biofilm formation on different materials for dental implants
Silva TSO, et al.
Journal of Visualized Experiments, e57756-e57756 (2018)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 97068-100ML-F | 04061833324899 |
| 97068-10ML-F-KC | 04061838078483 |



