Skip to Content
Merck

Skip To

90338

Triethylamine

ampule, ≥99.5% (GC)

Synonym(s):

N,N-Diethylethanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
UNSPSC Code:
12352116
NACRES:
NA.26
PubChem Substance ID:
EC Number:
204-469-4
Beilstein/REAXYS Number:
605283
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3.5 (vs air)

Quality Level

vapor pressure

51.75 mmHg ( 20 °C)

assay

≥99.5% (GC)

form

liquid

autoignition temp.

593 °F

quality

ampule

shelf life

42 mo.

expl. lim.

8 %

technique(s)

protein sequencing: suitable

impurities

≤0.1% water

refractive index

n20/D 1.401 (lit.)

pH

12.7 (15 °C, 100 g/L)

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Application

Conversion of the Beckman liquid phase sequencer to a gas-liquid phase sequencer[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
903401630490337
technique(s)

protein sequencing: suitable

technique(s)

-

technique(s)

-

technique(s)

-

assay

≥99.5% (GC)

assay

≥99.5% (GC)

assay

≥99% (GC)

assay

≥99.5% (GC)

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

liquid

form

liquid

form

liquid

form

liquid

shelf life

42 mo.

shelf life

36 mo.

shelf life

-

shelf life

42 mo.

density

0.726 g/mL at 25 °C (lit.)

density

0.727 g/mL at 20 °C, 0.726 g/mL at 25 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Triethylamine


signalword

Danger

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

flash_point_f

12.2 °F - closed cup

flash_point_c

-11 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A S Bhown et al.
Analytical biochemistry, 175(1), 39-51 (1988-11-15)
As an effective aid to extend the microsequencing capabilities the Beckman protein/peptide sequenator Series 890C has been successfully converted to a gas-liquid system, in which coupling buffer 25% trimethylamine was employed as a gas, and heptafluorobutyric acid as a liquid.
Kai Morino et al.
Scientific reports, 5, 8953-8953 (2015-05-21)
Well-trained clinicians may be able to provide diagnosis and prognosis from very short biomarker series using information and experience gained from previous patients. Although mathematical methods can potentially help clinicians to predict the progression of diseases, there is no method
Sumanta Kumar Padhi et al.
Inorganic chemistry, 51(15), 8091-8102 (2012-07-26)
Cyclometalated ruthenium complexes having C(^)N and N(^)C type coordinating ligands with NAD(+)/NADH function have been synthesized and characterized by spectroscopic methods. The variation of the coordinating position of σ-donating carbon atom leads to a drastic change in their properties. Both



Global Trade Item Number

SKUGTIN
90338-2ML04061838236203
90338-10X2ML04061833246702

Questions

Reviews

No rating value

Active Filters