This product is a solution containing approximately 50% polyethylene glycol 1000 in water.
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
Molecular Biology
Quality Level
product line
BioUltra
form
liquid
greener alternative product characteristics
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
concentration
~50% in H2O
impurities
DNases, none detected, RNases, none detected, insoluble matter, none detected, phosphatases, none detected, proteases, none detected
pH
6.0-7.5 (25 °C)
anion traces
chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg
cation traces
Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg
λ
neat
UV absorption
λ: 260 nm Amax: 0.6, λ: 280 nm Amax: 0.3
greener alternative category
SMILES string
C(CO)O
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
General description
Application
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form - | form liquid | form liquid |
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| impurities DNases, none detected, insoluble matter, none detected, proteases, none detected, RNases, none detected, phosphatases, none detected | impurities - | impurities - | impurities - |
| cation traces Al: ≤5 mg/kg, Ba: ≤5 mg/kg, Ca: ≤10 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, K: ≤200 mg/kg, Mg: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Sr: ≤5 mg/kg, As: ≤0.1 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Mn: ≤5 mg/kg, Pb: ≤5 mg/kg, Bi: ≤5 mg/kg, Li: ≤5 mg/kg, Zn: ≤5 mg/kg, Cr: ≤5 mg/kg, Na: ≤200 mg/kg | cation traces - | cation traces - | cation traces - |
| greener alternative product characteristics Safer Solvents and Auxiliaries | greener alternative product characteristics Safer Solvents and Auxiliaries | greener alternative product characteristics Safer Solvents and Auxiliaries | greener alternative product characteristics Safer Solvents and Auxiliaries |
| sustainability Greener Alternative Product | sustainability Greener Alternative Product | sustainability Greener Alternative Product | sustainability Greener Alternative Product |
Still not finding the right product?
Explore all of our products under Polyethylene glycol solution
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 76293-10ML-KC-F | 04061838085481 |
| 76293-100ML-F | 04061832922720 |
-
is this PEG 1000
1 answer-
Helpful?
-
