Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.25
UNSPSC Code:
41116107
Pricing and availability is not currently available.
grade
analytical standard
Quality Level
description
particle mass concentration 40 - 55 mg/mL
bp
212 °C
mp
32 °C
application(s)
glass & ceramic
industrial qc
pharmaceutical
format
neat
storage temp.
2-8°C
General description
Amino functionalized magnetic microparticles are a particle size standard, designed for characterizing particle size distribution (PSD) of test samples.
The product is superparamagnetic, comprising particles with iron oxide core and silane coating. BioMag® Plus Particles are approx. 1 micron in diameter.
The product is superparamagnetic, comprising particles with iron oxide core and silane coating. BioMag® Plus Particles are approx. 1 micron in diameter.
Application
Used for development and testing of particle size instruments.
Also used in microfluidic biosensors as a platform to immobilize bioactive materials for quantifying:
In addition, it finds application in electrochemical determination of drug residues in animal tissues.
Also used in microfluidic biosensors as a platform to immobilize bioactive materials for quantifying:
- mycotoxins in fruits[1] and feedstuffs
- emerging contaminants in river water samples
- fungal pathogens in fruits
In addition, it finds application in electrochemical determination of drug residues in animal tissues.
Features and Benefits
- suitable for routine instrument calibration checks, testing and corrections
- available in 10 mL pack size as a neat sample
Legal Information
BioMag is a registered trademark of Polysciences, Inc.
1 of 1
This Item | |||
|---|---|---|---|
| format neat | format neat | format neat | format neat |
| bp 212 °C | bp - | bp 212 °C | bp - |
| mp 32 °C | mp - | mp 32 °C | mp - |
| grade analytical standard | grade analytical standard | grade analytical standard | grade analytical standard |
| application(s) glass & ceramic | application(s) glass & ceramic | application(s) glass & ceramic | application(s) environmental |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Modified paramagnetic beads in a microfluidic system for the determination of zearalenone in feedstuffs samples
Panini NV, et al.
Food Chemistry, 125(2), 791-796 (2011)
Microfluidic immunosensor with micromagnetic beads coupled to carbon-based screen-printed electrodes (SPCEs) for determination of Botrytis cinerea in tissue of fruits
Fernandez-Baldo MA, et al.
Journal of Agricultural and Food Chemistry, 58(21), 11201-11206 (2010)
Nanostructured voltammetric sensor for ultra-trace anabolic drug determination in food safety field
Regiart M, et al.
Sensors and Actuators B, Chemical, 188, 1241-1249 (2013)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 53572-10ML-F | 04061837790607 |



