Skip to Content
Merck

Skip To

49449

L-Glutamic acid

≥99.5% (NT), BioUltra

Synonym(s):

(S)-2-Aminopentanedioic acid, Glu

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13
NACRES:
NA.26
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-293-7
MDL number:
Beilstein/REAXYS Number:
1723801
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Glutamic acid, BioUltra, ≥99.5% (NT)

product line

BioUltra

Quality Level

assay

≥99.5% (NT)

form

powder

optical activity

[α]20/D +31.5±1.0°, c = 5% in 5 M HCl

impurities

insoluble matter, passes filter test, ≤0.5% foreign amino acids

ign. residue

≤0.05% (as SO4)

loss

≤0.05% loss on drying, 20 °C (HV)

color

white

mp

205 °C (dec.) (lit.)

solubility

1 M HCl: 1 M at 20 °C, clear, colorless

density

1.54 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤100 mg/kg, sulfate (SO42-): ≤200 mg/kg

cation traces

Al: ≤5 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤200 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, NH4+: ≤100 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

1 M in 1 M HCl

UV absorption

λ: 260 nm Amax: 0.1, λ: 280 nm Amax: 0.1

application(s)

cell analysis

SMILES string

N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1

InChI key

WHUUTDBJXJRKMK-VKHMYHEASA-N

Biochem/physiol Actions

An excitatory amino acid neurotransmitter that is an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA).

Other Notes

Lesion-sparing agent, has excitotoxic properties[1]; May function as precipitating agent in protein crystallization[2]; Growth requirement of various microorganisms

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G8415W32850295436
form

powder

form

powder

form

powder or crystals

form

-

assay

≥99.5% (NT)

assay

98.5-100.5%

assay

98.5-101.5% dry basis (perchloric acid titration)

assay

-

product line

BioUltra

product line

-

product line

-

product line

TraceCERT®

solubility

1 M HCl: 1 M at 20 °C, clear, colorless

solubility

1 M HCl: 100 mg/mL

solubility

water: soluble 8.64 g/L at 25 °C

solubility

-

optical activity

[α]20/D +31.5±1.0°, c = 5% in 5 M HCl

optical activity

-

optical activity

[α]20/D +32°, c = 10 in 2 M HCl

optical activity

-

color

white

color

white

color

-

color

-


Still not finding the right product?

Explore all of our products under L-Glutamic acid


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



CRC Handbook of Microbiology, 4, 1-1 (1974)
Excitotoxic amino acids as neuroendocrine research tools.
J W Olney et al.
Methods in enzymology, 103, 379-393 (1983-01-01)
Protein crystallization: the growth of large-scale single crystals.
G L Gilliland et al.
Methods in enzymology, 104, 370-381 (1984-01-01)



Global Trade Item Number

SKUGTIN
49449-100G04061832410265
49449-1KG04061832410289
49449-500G04061832410326

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters