Skip to Content
Merck

Skip To

43819

DL-Dithiothreitol

≥99.0% (RT)

Synonym(s):

threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
UNSPSC Code:
12352201
NACRES:
NA.21
PubChem Substance ID:
EC Number:
222-468-7
Beilstein/REAXYS Number:
1719757
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥99.0% (RT)

form

powder

reaction suitability

reagent type: reductant

impurities

≤0.3% 4,5-dihydroxy-1,2-dithiane

mp

40-43 °C, 41-44 °C (lit.)

solubility

H2O: 50 mg/mL, clear, colorless, H2O: freely soluble, acetone: freely soluble, chloroform: freely soluble, diethyl ether: freely soluble, ethanol: freely soluble, ethyl acetate: freely soluble

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤250 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

Application

An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).
DL-Dithiothreitol is a pivotal component used in the preparation of buffer which in turn is utilized in the platelet cytosol preparation.[1] It is also important in isolation of RNA by using guanidinium salts and homogenisation of tissue[2] and cells[3].

Biochem/physiol Actions

DL-Dithiothreitol denatures proteins by reducing the disulphide bonds and reducing them to SH groups. It also prevents intramolecular and intermolecular disulfide bond formation between cysteine residues of proteins.[4]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0632D916343816
assay

≥99.0% (RT)

assay

≥98% (HPLC), ≥99.0% (titration)

assay

≥99%

assay

900-1100 mmol/L (Titration molarity)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

impurities

≤0.3% 4,5-dihydroxy-1,2-dithiane

impurities

-

impurities

-

impurities

DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected

mp

40-43 °C, 41-44 °C (lit.)

mp

41-44 °C (lit.)

mp

40-44 °C, 41-44 °C (lit.)

mp

-

form

powder

form

powder

form

powder

form

liquid


Still not finding the right product?

Explore all of our products under DL-Dithiothreitol


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



M W Radomski et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(13), 5193-5197 (1990-07-01)
Aggregation of human washed platelets with collagen is accompanied by a concentration-dependent increase in cyclic GMP but not cyclic AMP. NG-Monomethyl-L-arginine (L-MeArg), a selective inhibitor of nitric oxide (NO) synthesis from L-arginine, reduces this increase and enhances aggregation. L-Arginine, which
R G Knowles et al.
Proceedings of the National Academy of Sciences of the United States of America, 86(13), 5159-5162 (1989-07-01)
A soluble enzyme obtained from rat forebrain catalyzes the NADPH-dependent formation of nitric oxide (NO) and citrulline from L-arginine. The NO formed stimulates the soluble guanylate cyclase and this stimulation is abolished by low concentrations of hemoglobin. The synthesis of
M B Donati et al.
Cancer research, 46(12 Pt 1), 6471-6474 (1986-12-01)
It has repeatedly been proposed that fibrin plays a role in tumor growth and metastasis. Among tumor cell products or activities which may promote clot formation, cancer procoagulant (CP), a direct activator of coagulation factor X, has been suggested to



Global Trade Item Number

SKUGTIN
43819-25G04061832132938
43819-5G04061832132990
43819-1G04061835515509

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters